Tempus™ 12-Port RNA Isolation Kit - FAQs
Tempus™ 12-Port RNA Isolation Kit - FAQs
View additional product information for Tempus™ 12-Port RNA Isolation Kit - FAQs (4378672)
1 product FAQs found
I am doing blood work and using the Tempus collection tubes. Is the Tempus system compatible with the RiboPure-Blood RNA Purification Kit?
No, please follow the RNA isolation methods described with the Tempus system. The collection tubes are not compatible with the RiboPure kit.