Product Image
Applied Biosystems™

TaqMan™ Array Mouse Endogenous Control Panel

The Applied Biosystems™ TaqMan™ Mouse Endogenous Control Array is a 384-well micro flluidic card containing 16 mouse TaqMan™ Gene Expression자세히 알아보기
Have Questions?
카탈로그 번호수량
43787022 cards
카탈로그 번호 4378702
제품 가격(KRW)
1,156,000
온라인 행사
Ends: 31-Dec-2026
1,445,000
할인액 289,000 (20%)
Each
수량:
2 cards
제품 가격(KRW)
1,156,000
온라인 행사
Ends: 31-Dec-2026
1,445,000
할인액 289,000 (20%)
Each
The Applied Biosystems™ TaqMan™ Mouse Endogenous Control Array is a 384-well micro flluidic card containing 16 mouse TaqMan™ Gene Expression Assays commonly used as endogenous controls arrayed in triplicates. This product is compatible with an upgraded Applied Biosystems 7900HT Fast Real-Time PCR System.

The array is pre-formatted and inventoried, cost-effective, convenient and easy to use without needing expensive robotics. Results are reproducible and consistent across samples, studies, and labs —the same data quality from card-to-card and lot-to-lot — even with different operators.
For Research Use Only. Not for use in diagnostic procedures.
사양
형태Dried Down
포함Control Primer-Probe Set(s)
내부 프로브 수정MGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5')
라벨 또는 염료FAM
제품라인TaqMan
수량2 cards
반응 속도Standard
타겟 유전자Housekeeping & Control Gene Panel
유형Array Mouse Endogenous Control Panel
형식384-well Microfluidic Card
Mouse
Unit SizeEach
구성 및 보관
TaqMan™ Mouse Endogenous Control Array is sold in a pack of 2 cards and include a CD with assay information and an instrument software input file. The cards should be stored at 4°C.

자주 묻는 질문(FAQ)

What are the storage recommendations for TaqMan Array Cards (TAC)?

We recommend storing TaqMan Array Cards (TAC) at 2-8 degrees C. TaqMan Array Cards (TAC) are stable for 2 years from the manufacturing date. The expiration date is listed on the packaging.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

Can poly A+ purified RNA samples be used with the TaqMan Human Endogenous Control Plate (Cta. No. 4309199)?

No. The plate has been designed to evaluate only total RNA. Ribosomal RNA will be removed from poly A+ purified RNA samples, and is required for accurate detection of the 18s rRNA endogenous control assay used on the plate.