Do you need this panel in a 384-well plate? Please visit Specialty Plating to learn more.
Applied Biosystems™
TaqMan™ Array Human Phosphodiesterase Panel
The Applied Biosystems™ TaqMan™ Human Phosphodiesterase Array is an efficient, easy-to-use micro fluidic card for quantitative gene expression analysis ofRead more
Have Questions?
Catalog Number
Quantity
4378705
4 cards
Catalog number 4378705
Price (USD)
2,635.00
Each
Quantity:
4 cards
Price (USD)
2,635.00
Each
The Applied Biosystems™ TaqMan™ Human Phosphodiesterase Array is an efficient, easy-to-use micro fluidic card for quantitative gene expression analysis of inflammation genes important in drug discovery. This product is designed with 48 TaqMan™ Gene Expression Assays arrayed in 384 wells and is compatible with an upgraded Applied Biosystems 7900HT Fast Real-Time PCR System and sold in packs of 4. The array contains 43 human assays and five human controls. Orthologous genes to the human panel are in the Rat Phosphodiesterase Panel with two exceptions: Pde6g and Gdpd4 are missing and two added controls (Arbp and Ywhaz) are in the rat panel.
The array is pre-formatted and inventoried, cost-effective, convenient and easy to use without needing expensive robotics. Results are reproducible and consistent across samples, studies, and labs —the same data quality from card-to-card and lot-to-lot — even with different operators.
For Research Use Only. Not for use in diagnostic procedures.
Research CategoryCell Cycle & Proliferation, Neurobiology, Toxicology & Drug Metabolism, Immunology, Oncology
Target GeneIncludes five distinct phosphodiesterase families
TypeArray Human Phosphodiesterase Panel
Format384-well Microfluidic Card
SpeciesHuman
Unit SizeEach
Contents & Storage
Each TaqMan™ Array well contains a single TaqMan™ Assay for one 2 μL reaction. The cards should be stored at 4°C.
Frequently asked questions (FAQs)
What are the storage recommendations for TaqMan Array Cards (TAC)?
We recommend storing TaqMan Array Cards (TAC) at 2-8 degrees C. TaqMan Array Cards (TAC) are stable for 2 years from the manufacturing date. The expiration date is listed on the packaging.