Not what you're looking for? Search all of our TaqMan® arrays for your gene or pathway of interest.
Do you need this panel in a 384-well plate? Please visit Specialty Plating to learn more.
Product Image
Applied Biosystems™

TaqMan™ Array Mouse Alzheimer's Panel

The Applied Biosystems™ TaqMan™ Mouse Alzheimer's Array is a 384-well micro flluidic card containing 96 human TaqMan™ Gene Expression Assays.Read more
Have Questions?
Catalog NumberQuantity
43787144 cards
Catalog number 4378714
Price (USD)
2,635.00
Each
Quantity:
4 cards
Price (USD)
2,635.00
Each
The Applied Biosystems™ TaqMan™ Mouse Alzheimer's Array is a 384-well micro flluidic card containing 96 human TaqMan™ Gene Expression Assays. This product is compatible with an upgraded Applied Biosystems 7900HT Fast Real-Time PCR System and sold in packs of 4.

TaqMan™ Arrays for Alzheimer’s for mouse closely follow the human panel and are from orthologous genes of the human assays. The mouse panel has 92 assays and four controls (18S, actb, hprt1 and ipo8).The array is pre-formatted and inventoried, cost-effective, convenient and easy to use without needing expensive robotics. Results are reproducible and consistent across samples, studies, and labs —the same data quality from card-to-card and lot-to-lot — even with different operators.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormDried Down
IncludesTaqMan Assay for One 2 uL Reaction
Internal Probe ModificationMGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5')
Label or DyeFAM
Product LineTaqMan
Quantity4 cards
Reaction SpeedStandard
Research CategoryNeurobiology
Target Gene94 assays for APP processing, cholesterol biosynthesis, and other Alzheimer's targets
TypeArray Mouse Alzheimer's Panel
Format384-well Microfluidic Card
SpeciesMouse
Unit SizeEach
Contents & Storage
Each TaqMan™ Array well contains a single TaqMan™ Assay for one 2 μL reaction. The cards should be stored at 4°C.

Frequently asked questions (FAQs)

What are the storage recommendations for TaqMan Array Cards (TAC)?

We recommend storing TaqMan Array Cards (TAC) at 2-8 degrees C. TaqMan Array Cards (TAC) are stable for 2 years from the manufacturing date. The expiration date is listed on the packaging.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.