7500 Fast Upgrade - Citations

7500 Fast Upgrade - Citations

View additional product information for 7500 Fast Upgrade - Citations (4379638)

Showing 100 of 842 total product Citations

Citations & References
Abstract
Characterization of matrix metalloproteinase-2 and matrix metalloproteinase-9 and their inhibitors in equine granulosa cells in vivo and in vitro
AuthorsSessions, DR; Vick, MM; Fitzgerald, BP
JournalJOURNAL OF ANIMAL SCIENCE
PubMed ID
Matrix metalloproteinases (MMP) and tissue inhibitors of MMP (TIMP) regulate tissue remodeling events necessary for ovulation. Thus, changes in MMP and TIMP expression and protein enzyme activity were examined in vivo and in vitro during follicular development and atresia in the horse. Equine granulosa cells and follicular fluid from medium ... More
Characterization of matrix metalloproteinase-2 and matrix metalloproteinase-9 and their inhibitors in equine granulosa cells in vivo and in vitro
AuthorsSessions, DR; Vick, MM; Fitzgerald, BP
JournalJOURNAL OF ANIMAL SCIENCE
PubMed ID
Matrix metalloproteinases (MMP) and tissue inhibitors of MMP (TIMP) regulate tissue remodeling events necessary for ovulation. Thus, changes in MMP and TIMP expression and protein enzyme activity were examined in vivo and in vitro during follicular development and atresia in the horse. Equine granulosa cells and follicular fluid from medium ... More
Molecular cloning and characterization of porcine calcineurin-alpha subunit expression in skeletal muscle.
AuthorsDepreux FF, Scheffler JM, Grant AL, Bidwell CA, Gerrard DE
JournalJ Anim Sci
PubMed ID19897633
The calmodulin/Ca(2+)-dependent serine/threonine phophatase, calcineurin (CaN), has been implicated in controlling muscle fiber phenotype. However, little information is available concerning the expression of CaN in porcine skeletal muscle. Therefore, the porcine CaN alpha (CaN-A) was cloned by reverse transcription-PCR and its expression characterized in selected porcine skeletal muscles. We successfully ... More
Genetic analysis of hepatitis C virus with defective genome and its infectivity in vitro.
AuthorsSugiyama K, Suzuki K, Nakazawa T, Funami K, Hishiki T, Ogawa K, Saito S, Shimotohno KW, Suzuki T, Shimizu Y, Tobita R, Hijikata M, Takaku H, Shimotohno K
JournalJ Virol
PubMed ID19369330
Replication and infectivity of hepatitis C virus (HCV) with a defective genome is ambiguous. We molecularly cloned 38 HCV isolates with defective genomes from 18 patient sera. The structural regions were widely deleted, with the 5' untranslated, core, and NS3-NS5B regions preserved. All of the deletions were in frame, ... More
Cell lines as candidate reference materials for quality control of ERBB2 amplification and expression assays in breast cancer.
AuthorsXiao Y, Gao X, Maragh S, Telford WG, Tona A
JournalClin Chem
PubMed ID19443566
BACKGROUND: Human epidermal growth factor receptor 2 (HER2) is an important biomarker whose status plays a pivotal role in therapeutic decision-making for breast cancer patients and in determining their clinical outcomes. Ensuring the accuracy and reproducibility of HER2 assays by immunohistochemistry (IHC) and by fluorescence in situ hybridization (FISH) ... More
HIV-1 transactivator protein induction of suppressor of cytokine signaling-2 contributes to dysregulation of IFN{gamma} signaling.
AuthorsCheng SM, Li JC, Lin SS, Lee DC, Liu L, Chen Z, Lau AS
JournalBlood
PubMed ID19279332
HIV infection remains a worldwide threat. HIV-1 transactivator protein Tat is one of the retroviral proteins identified as a key immunomodulator in AIDS pathogenesis. Although the primary function of Tat is to regulate HIV-1 replication in the infected cell, it also dysregulates cytokine production resulting in perturbation of the host ... More
Dynamic PolyConjugates for targeted in vivo delivery of siRNA to hepatocytes.
AuthorsRozema DB, Lewis DL, Wakefield DH, Wong SC, Klein JJ, Roesch PL, Bertin SL, Reppen TW, Chu Q, Blokhin AV, Hagstrom JE, Wolff JA
JournalProc Natl Acad Sci U S A
PubMed ID17652171
Achieving efficient in vivo delivery of siRNA to the appropriate target cell would be a major advance in the use of RNAi in gene function studies and as a therapeutic modality. Hepatocytes, the key parenchymal cells of the liver, are a particularly attractive target cell type for siRNA delivery ... More
Hepatocyte growth factor/scatter factor promotes retinal angiogenesis through increased urokinase expression.
AuthorsColombo ES, Menicucci G, McGuire PG, Das A
JournalInvest Ophthalmol Vis Sci
PubMed ID17389513
PURPOSE: The purpose of this study was to determine the role of hepatocyte growth factor (HGF) and c-Met in the initiation and development of retinal neovascularization and to determine whether inhibition of this system can suppress the extent of angiogenesis in an animal model. METHODS: Retinal tissues from animals ... More
A novel one-armed anti-c-Met antibody inhibits glioblastoma growth in vivo.
AuthorsMartens T, Schmidt NO, Eckerich C, Fillbrandt R, Merchant M, Schwall R, Westphal M, Lamszus K
JournalClin Cancer Res
PubMed ID17062691
PURPOSE: Expression of the receptor tyrosine kinase c-Met and its ligand scatter factor/hepatocyte growth factor (SF/HGF) are strongly increased in glioblastomas, where they promote tumor proliferation, migration, invasion, and angiogenesis. We used a novel one-armed anti-c-Met antibody to inhibit glioblastoma growth in vivo. EXPERIMENTAL DESIGN: U87 glioblastoma cells (c-Met ... More
Breed difference and regulation of the porcine adipose triglyceride lipase and hormone sensitive lipase by TNFα
AuthorsShan, T; Wu, T; Reng, Y; Wang, Y
JournalANIMAL GENETICS
PubMed ID
Adipose triglyceride lipase (ATGL) and hormone sensitive lipase (HSL) are major novel triglyceride lipases in animals. The aim of this study was to determine if there are differences in the porcine ATGL (pATGL) and HSL genes between Jinhua pigs (a fatty breed) and Landrace pigs (a leaner breed). In addition, ... More
Neuropathogenic and non-neuropathogenic genotypes of Equid Herpesvirus type 1 in Argentina
AuthorsVissani, MA; Becerra, ML; Perglione, CO; Tordoya, MS; Mino, S; Barrandeguy, M
JournalVETERINARY MICROBIOLOGY
PubMed ID
Infection with Equid Herpesvirus type 1 (EHV-1) leads to respiratory disease, abortion, and neurological disorders in horses. Molecular epidemiology studies have demonstrated that a single nucleotide polymorphism (A(2254)/G(2254)) in the genome region of the open reading frame 30 (ORF30), which results in an amino acid variation (N(752)/D(752)) of the EHV-1 ... More
Expression profiling and electrophysiological studies suggest a major role for Orai1 in the store-operated Ca(2+) influx pathway of platelets and megakaryocytes
AuthorsTolhurst, G; Carter, RN; Amisten, S; Holdich, JP; Erlinge, D; Mahaut-Smith, MP
JournalPLATELETS
PubMed ID
Store-operated Ca(2+) influx represents a major route by which cytosolic Ca(2+) can be elevated during platelet activation, yet its molecular identity in this cell type remains highly controversial. Using quantitative RT-PCR analysis of candidate receptor-operated cation entry pathways in human platelets, we show a 30-fold higher expression of message for ... More
Retinoic acid modulates chromatin to potentiate tumor necrosis factor alpha signaling on the DIF2 promoter.
AuthorsWitcher M, Pettersson F, Dupere-Richer D, Padovani A, Summers-Deluca L, Baldwin AS, Miller WH Jr
JournalNucleic Acids Res
PubMed ID18039708
'Transcriptional activation by nuclear hormone receptors is well characterized, but their cooperation with other signaling pathways to activate transcription remains poorly understood. Tumor necrosis factor alpha (TNFalpha) and all-trans retinoic acid (RA) induce monocytic differentiation of acute promyelocytic leukemia (APL) cells in a synergistic manner. We used the promoter of ... More
Clinically distinct epigenetic subgroups in Silver-Russell syndrome: the degree of H19 hypomethylation associates with phenotype severity and genital and skeletal anomalies.
AuthorsBruce S, Hannula-Jouppi K, Peltonen J, Kere J, Lipsanen-Nyman M
JournalJ Clin Endocrinol Metab
PubMed ID19017756
'CONTEXT: The H19 imprinting control region (ICR), located on chromosome 11p15.5, has been reported hypomethylated in 20-65% of Silver-Russell syndrome (SRS) patients. OBJECTIVE: We investigated the methylation status of 11p15.5 ICRs in SRS patients and children born small for gestational age (SGA) to clarify the relationship between phenotype and ... More
CNTNAP2 and NRXN1 Are Mutated in Autosomal-Recessive Pitt-Hopkins-like Mental Retardation and Determine the Level of a Common Synaptic Protein in Drosophila
AuthorsZweier, C; de Jong, EK; Zweier, M; Orrico, A; Ousager, LB; Collins, AL; Bijlsma, EK; Oortveld, MAW; Ekici, AB; Reis, A; Schenck, A; Rauch, A
Journal
PubMed ID
'Heterozygous copy-number variants and SNPs of CNTNAP2 and NRXN1, two distantly related members of the neurexin superfamily, have been repeatedly associated with a wide spectrum of neuropsychiatric disorders, such as developmental language disorders, autism spectrum disorders, epilepsy, and schizophrenia. We now identified homozygous and compound-heterozygous deletions and mutations via molecular ... More
Blockade of the ubiquitin protease UBP43 destabilizes transcription factor PML/RARalpha and inhibits the growth of acute promyelocytic leukemia.
AuthorsGuo Y, Dolinko AV, Chinyengetere F, Stanton B, Bomberger JM, Demidenko E, Zhou DC, Gallagher R, Ma T, Galimberti F, Liu X, Sekula D, Freemantle S, Dmitrovsky E
JournalCancer Res
PubMed ID20935222
'More effective treatments for acute promyelocytic leukemia (APL) are needed. APL cell treatment with all-trans-retinoic acid (RA) degrades the chimeric, dominant-negative-acting transcription factor promyelocytic leukemia gene (PML)/RARalpha, which is generated in APL by chromosomal translocation. The E1-like ubiquitin-activating enzyme (UBE1L) associates with interferon-stimulated gene ISG15 that binds and represses PML/RARalpha ... More
Inhibition of cyclooxygenase-2 suppresses lymph node metastasis via reduction of lymphangiogenesis.
AuthorsIwata C, Kano MR, Komuro A, Oka M, Kiyono K, Johansson E, Morishita Y, Yashiro M, Hirakawa K, Kaminishi M, Miyazono K
JournalCancer Res
PubMed ID17974958
'Cyclooxygenase-2 (COX-2) inhibitor has been reported to suppress tumor progression. However, it is unclear whether this inhibitor can also prevent lymphatic metastasis. To determine the effects of COX-2 inhibitor on lymphatic metastasis, etodolac, a COX-2 inhibitor, was given p.o. to mice bearing orthotopic xenografts or with carcinomatous peritonitis induced with ... More
Sirtuin 1 is required for antagonist-induced transcriptional repression of androgen-responsive genes by the androgen receptor.
AuthorsDai Y, Ngo D, Forman LW, Qin DC, Jacob J, Faller DV
JournalMol Endocrinol
PubMed ID17505061
'Androgen antagonists or androgen deprivation is a primary therapeutic modality for the treatment of prostate cancer. Invariably, however, the disease becomes progressive and unresponsive to androgen ablation therapy (hormone refractory). The molecular mechanisms by which the androgen antagonists inhibit prostate cancer proliferation are not fully defined. In this report, we ... More
MicroRNA-223 regulates Glut4 expression and cardiomyocyte glucose metabolism.
AuthorsLu H, Buchan RJ, Cook SA
JournalCardiovasc Res
PubMed ID20080987
'AIMS: MicroRNAs (miRNAs) are important for cardiac function and tissue metabolism. The aim of the present study is to investigate the role(s) of miRNAs in the insulin-resistant heart. METHODS AND RESULTS: Left ventricular biopsies were collected from patients with or without type 2 diabetes and from patients with left ... More
Identification of immune parameters to differentiate disease states among sheep infected with Mycobacterium avium subsp. paratuberculosis.
AuthorsGillan S, O'Brien R, Hughes AD, Griffin JF
JournalClin Vaccine Immunol
PubMed ID19923568
'Johne''s disease, a chronic enteritis of ruminants, is caused by infection with Mycobacterium avium subsp. paratuberculosis. Three distinct forms have been observed in sheep: paucibacillary disease (PB), multibacillary disease (MB), and asymptomatic infection (AS). In this study, immune parameters for animals naturally infected with M. avium subsp. paratuberculosis and ... More
Dependence of stress resistance on a spore coat heteropolysaccharide in Dictyostelium.
AuthorsWest CM, Nguyen P, van der Wel H, Metcalf T, Sweeney KR, Blader IJ, Erdos GW
JournalEukaryot Cell
PubMed ID18996984
'In Dictyostelium, sporulation occurs synchronously as prespore cells approach the apex of the aerial stalk during culmination. Each prespore cell becomes surrounded by its own coat comprised of a core of crystalline cellulose and a branched heteropolysaccharide sandwiched between heterogeneous cysteine-rich glycoproteins. The function of the heteropolysaccharide, which consists of ... More
Thyroid transcription factor-1 inhibits transforming growth factor-beta-mediated epithelial-to-mesenchymal transition in lung adenocarcinoma cells.
AuthorsSaito RA, Watabe T, Horiguchi K, Kohyama T, Saitoh M, Nagase T, Miyazono K
JournalCancer Res
PubMed ID19293183
'Thyroid transcription factor-1 (TTF-1) is expressed in lung cancer, but its functional roles remain unexplored. TTF-1 gene amplification has been discovered in a part of lung adenocarcinomas, and its action as a lineage-specific oncogene is highlighted. Epithelial-to-mesenchymal transition (EMT) is a crucial event for cancer cells to acquire ... More
Prevalence of t(12;21)[ETV6-RUNX1]-positive cells in healthy neonates.
AuthorsLausten-Thomsen U, Madsen HO, Vestergaard TR, Hjalgrim H, Nersting J, Schmiegelow K
JournalBlood
PubMed ID20713965
't(12;21)(p13;q22)[ETV6-RUNX1] is the most common chromosomal translocation in childhood acute lymphoblastic leukemia, and it can often be backtracked to Guthrie cards supporting prenatal initiation and high levels of circulating t(12;21)-positive cells at birth. To explore the prevalence of ETV6-RUNX1-positive cells in healthy neonates, mononuclear cells from 1417 umbilical cord blood ... More
Development and validation of a multiplex real-time PCR assay for simultaneous genotyping and human T-lymphotropic virus type 1, 2, and 3 proviral load determination.
AuthorsMoens B, Lopez G, Adaui V, Gonzalez E, Kerremans L, Clark D, Verdonck K, Gotuzzo E, Vanham G, Cassar O, Gessain A, Vandamme AM, Van Dooren S
JournalJ Clin Microbiol
PubMed ID19741085
'The human T-lymphotropic virus (HTLV) proviral load remains the best surrogate marker for disease progression. Real-time PCR techniques have been developed for detection and quantification of cosmopolitan HTLV type 1a (HTLV-1a) and HTLV-2. Since a growing level of diversity in subtypes and genotypes is observed, we developed a multiplex ... More
von Hippel-Lindau protein regulates transition from the fetal to the adult circulatory system in retina.
AuthorsKurihara T, Kubota Y, Ozawa Y, Takubo K, Noda K, Simon MC, Johnson RS, Suematsu M, Tsubota K, Ishida S, Goda N, Suda T, Okano H
JournalDevelopment
PubMed ID20388654
'In early neonates, the fetal circulatory system undergoes dramatic transition to the adult circulatory system. Normally, embryonic connecting vessels, such as the ductus arteriosus and the foramen ovale, close and regress. In the neonatal retina, hyaloid vessels maintaining blood flow in the embryonic retina regress, and retinal vessels take over ... More
Activation of paternally expressed genes and perinatal death caused by deletion of the Gtl2 gene.
AuthorsZhou Y, Cheunsuchon P, Nakayama Y, Lawlor MW, Zhong Y, Rice KA, Zhang L, Zhang X, Gordon FE, Lidov HG, Bronson RT, Klibanski A
JournalDevelopment
PubMed ID20610486
'The Dlk1-Gtl2 imprinting locus is located on mouse distal chromosome 12 and consists of multiple maternally expressed non-coding RNAs and several paternally expressed protein-coding genes. The imprinting of this locus plays a crucial role in embryonic development and postnatal growth. At least one cis-element, the intergenic differentially methylated region (IG-DMR) ... More
Genome-wide analysis of mRNA targets for Caenorhabditis elegans FBF, a conserved stem cell regulator.
AuthorsKershner AM, Kimble J
JournalProc Natl Acad Sci U S A
PubMed ID20142496
'Stem cells are essential for tissue generation during the development of multicellular creatures, and for tissue homeostasis in adults. The great therapeutic promise of stem cells makes understanding their regulation a high priority. PUF RNA-binding proteins have a conserved role in promoting self-renewal of germline stem cells. Here we use ... More
Profiling microRNA expression in hepatocellular carcinoma reveals microRNA-224 up-regulation and apoptosis inhibitor-5 as a microRNA-224-specific target.
AuthorsWang Y, Lee AT, Ma JZ, Wang J, Ren J, Yang Y, Tantoso E, Li KB, Ooi LL, Tan P, Lee CG
JournalJ Biol Chem
PubMed ID18319255
'Like other cancers, aberrant gene regulation features significantly in hepatocellular carcinoma (HCC). MicroRNAs (miRNAs) were recently found to regulate gene expression at the post-transcriptional/translational levels. The expression profiles of 157 miRNAs were examined in 19 HCC patients, and 19 up-regulated and 3 down-regulated miRNAs were found to be associated with ... More
Analysis of tetracycline resistance tet(W) genes and their flanking sequences in intestinal Bifidobacterium species.
AuthorsAmmor MS, Florez AB, Alvarez-Martin P, Margolles A, Mayo B
JournalJ Antimicrob Chemother
PubMed ID18614524
'OBJECTIVES: The tet(W) gene provides tetracycline resistance to a wide range of anaerobic intestinal and ruminal bacteria, but little is known about the molecular organization of the tet(W) gene. The aim of this study was to gain new insights into the molecular organization of the tet(W) gene in bifidobacteria ... More
Nuclear cathepsin F regulates activation markers in rat hepatic stellate cells.
AuthorsMaubach G, Lim MC, Zhuo L
JournalMol Biol Cell
PubMed ID18667530
'Activation of hepatic stellate cells during liver fibrosis is a major event facilitating an increase in extracellular matrix deposition. The up-regulation of smooth muscle alpha-actin and collagen type I is indicative of the activation process. The involvement of cysteine cathepsins, a class of lysosomal cysteine proteases, has not been studied ... More
Long-term administration of estradiol decreases expression of hepatic lipogenic genes and improves insulin sensitivity in ob/ob mice: a possible mechanism is through direct regulation of signal transducer and activator of transcription 3.
AuthorsGao H, Bryzgalova G, Hedman E, Khan A, Efendic S, Gustafsson JA, Dahlman-Wright K
JournalMol Endocrinol
PubMed ID16627594
'In this study, we used ob/ob mice as a model to investigate the effects of long-term estradiol administration on insulin sensitivity and to explore the mechanisms that underlie the antidiabetic effects of estrogen on mouse liver. Female ob/ob mice were randomly divided into two groups and given estradiol (100  ... More
Differential regulation of dopaminergic gene expression by Er81.
AuthorsCave JW, Akiba Y, Banerjee K, Bhosle S, Berlin R, Baker H
JournalJ Neurosci
PubMed ID20357122
'A recent study proposed that differentiation of dopaminergic neurons requires a conserved "dopamine motif" (DA-motif) that functions as a binding site for ETS DNA binding domain transcription factors. In the mammalian olfactory bulb (OB), the expression of a set of five genes [including tyrosine hydroxylase (Th)] that are necessary for ... More
Interleukin-10 attenuates vascular responses to endothelin-1 via effects on ERK1/2-dependent pathway.
AuthorsGiachini FR, Zemse SM, Carneiro FS, Lima VV, Carneiro ZN, Callera GE, Ergul A, Webb RC, Tostes RC
JournalAm J Physiol Heart Circ Physiol
PubMed ID19074677
'Interleukin-10 (IL-10) is an anti-inflammatory cytokine with protective actions on the vasculature. On the other hand, endothelin (ET)-1 has potent vasoconstrictor, mitogenic, and proinflammatory activities, which have been implicated in the pathophysiology of a number of cardiovascular diseases. We hypothesized that, in a condition where ET-1 expression is upregulated, i.e., ... More
Environmentally relevant concentrations of 17alpha-ethinylestradiol (EE2) interfere with the growth hormone (GH)/insulin-like growth factor (IGF)-I system in developing bony fish.
AuthorsShved N, Berishvili G, Baroiller JF, Segner H, Reinecke M
JournalToxicol Sci
PubMed ID18660547
'The aim of this study was to evaluate whether effects of environmental estrogens on fish growth and reproduction may be mediated via modulating the growth hormone (GH)/insulin-like growth factor I (IGF-I) system. To this end, developing male and female monosex populations of tilapia were exposed to 17alpha-ethinylestradiol (EE2) at ... More
Expression of hepatitis C virus (HCV) structural proteins in trans facilitates encapsidation and transmission of HCV subgenomic RNA.
AuthorsAdair R, Patel AH, Corless L, Griffin S, Rowlands DJ, McCormick CJ
JournalJ Gen Virol
PubMed ID19223490
'A characteristic of many positive-strand RNA viruses is that, whilst replication of the viral genome is dependent on the expression of the majority of non-structural proteins in cis, virus particle formation can occur when most or all of the structural proteins are co-expressed in trans. Making use of a ... More
Suppressor of cytokine signalling-3 is up-regulated by androgen in prostate cancer cell lines and inhibits androgen-mediated proliferation and secretion.
AuthorsNeuwirt H, Puhr M, Cavarretta IT, Mitterberger M, Hobisch A, Culig Z
JournalEndocr Relat Cancer
PubMed ID18045952
'Suppressors of cytokine signalling (SOCS) are induced by interleukins (ILs) and various peptide hormones and may prevent sustained activation of signalling pathways. We have previously shown that SOCS-3 antagonizes regulation of cellular events by cAMP and is expressed in human prostate cancer. To investigate possible effects of androgen on SOCS-3 ... More
Generation of mice transgenic for human CYP2C18 and CYP2C19: characterization of the sexually dimorphic gene and enzyme expression.
AuthorsLofgren S, Baldwin RM, Hiratsuka M, Lindqvist A, Carlberg A, Sim SC, Schulke M, Snait M, Edenro A, Fransson-Steen R, Terelius Y, Ingelman-Sundberg M
JournalDrug Metab Dispos
PubMed ID18276835
'CYP2C19 is an important enzyme for human drug metabolism, and it also participates in the metabolism of endogenous substrates, whereas the CYP2C18 enzyme is not expressed in human liver despite high mRNA expression. Mice transgenic for the human CYP2C18 and CYP2C19 genes were generated. Quantitative mRNA analysis showed CYP2C18 and ... More
Phosphatidylinositol 4-kinase III beta is a target of enviroxime-like compounds for antipoliovirus activity.
AuthorsArita M, Kojima H, Nagano T, Okabe T, Wakita T, Shimizu H
JournalJ Virol
PubMed ID21177810
'Enviroxime is an antienterovirus compound that targets viral protein 3A and/or 3AB and suppresses a step in enterovirus replication by unknown mechanism. To date, four antienterovirus compounds, i.e., GW5074, Flt3 inhibitor II, TTP-8307, and AN-12-H5, are known to have similar mutations in the 3A protein-encoding region causing resistance to enviroxime ... More
Regulation of soluble guanylyl cyclase-alpha1 expression in chronic hypoxia-induced pulmonary hypertension: role of NFATc3 and HuR.
Authorsde Frutos S, Nitta CH, Caldwell E, Friedman J, Gonzalez Bosc LV
JournalAm J Physiol Lung Cell Mol Physiol
PubMed ID19592461
'The nitric oxide/soluble guanylyl cyclase (sGC) signal transduction pathway plays an important role in smooth muscle relaxation and phenotypic regulation. However, the transcriptional regulation of sGC gene expression is largely unknown. It has been shown that sGC expression increases in pulmonary arteries from chronic hypoxia-induced pulmonary hypertensive animals. Since ... More
DNA methylation and histone modifications cause silencing of Wnt antagonist gene in human renal cell carcinoma cell lines
AuthorsKawamoto, K; Hirata, H; Kikuno, N; Tanaka, Y; Nakagawa, M; Dahiya, R
Journal
PubMed ID
'Secreted frizzled-related protein 2 (sFRP2) is a negative modulator of the Wingless-type (Wnt) signaling pathway, and shown to be inactivated in renal cell carcinoma (RCC). However, the molecular mechanism of silencing of sFRP2 is not fully understood. Our study was designed to elucidate the silencing mechanism of sFRP2 in RCC. ... More
Gene expression profiling defines ATP as a key regulator of human dendritic cell functions.
AuthorsBles N, Horckmans M, Lefort A, Libert F, Macours P, El Housni H, Marteau F, Boeynaems JM, Communi D
JournalJ Immunol
PubMed ID17785789
'Extracellular ATP and PGE2 are two cAMP-elevating agents inducing semimaturation of human monocyte-derived dendritic cells (MoDCs). We have extensively compared the gene expression profiles induced by adenosine 5''-O-(3-thiotriphosphate) (ATPgammaS) and PGE2 in human MoDCs using microarray technology. At 6 h of stimulation, ATPgammaS initiated an impressive expression profile compared ... More
Differential expression of microRNA-675, microRNA-139-3p and microRNA-335 in benign and malignant adrenocortical tumours.
AuthorsSchmitz KJ, Helwig J, Bertram S, Sheu SY, Suttorp AC, Seggewiss J, Willscher E, Walz MK, Worm K, Schmid KW
JournalJ Clin Pathol
PubMed ID21471143
'Background For the clinical management of adrenocortical neoplasms it is crucial to correctly distinguish between benign and malignant tumours. Even histomorphologically based scoring systems do not allow precise separation in single lesions, thus novel parameters are desired which offer a more accurate differentiation. The tremendous potential of microRNAs (miRNAs) as ... More
Activation of Macrophages by Polysaccharide-protein Complex from Lycium barbarum L.
AuthorsChen, ZS; Soo, MY; Srinivasan, N; Tan, BKH; Chan, SH
Journal
PubMed ID
'Macrophages play crucial roles in innate immunity. This paper reports that a polysaccharide-protein complex isolated from Lycium barbarum (LBP) is able to activate macrophages. LBP was isolated from Lycium barbarum fruit and separated to five homogenous fractions, designated LBPF1, LBPF2, LBPF3, LBPF4 and LBPF5. It was found that LBP (50 ... More
Mapping accessible chromatin regions using Sono-Seq.
AuthorsAuerbach RK, Euskirchen G, Rozowsky J, Lamarre-Vincent N, Moqtaderi Z, Lefrancois P, Struhl K, Gerstein M, Snyder M
JournalProc Natl Acad Sci U S A
PubMed ID19706456
'Disruptions in local chromatin structure often indicate features of biological interest such as regulatory regions. We find that sonication of cross-linked chromatin, when combined with a size-selection step and massively parallel short-read sequencing, can be used as a method (Sono-Seq) to map locations of high chromatin accessibility in promoter regions. ... More
A gain-of-function mutation of Arabidopsis cryptochrome1 promotes flowering.
AuthorsExner V, Alexandre C, Rosenfeldt G, Alfarano P, Nater M, Caflisch A, Gruissem W, Batschauer A, Hennig L
JournalPlant Physiol
PubMed ID20926618
'Plants use different classes of photoreceptors to collect information about their light environment. Cryptochromes are blue light photoreceptors that control deetiolation, entrain the circadian clock, and are involved in flowering time control. Here, we describe the cry1-L407F allele of Arabidopsis (Arabidopsis thaliana), which encodes a hypersensitive cryptochrome1 (cry1) protein. Plants ... More
Hypoglycemia from IGF2 overexpression associated with activation of fetal promoters and loss of imprinting in a metastatic hemangiopericytoma.
AuthorsLawson EA, Zhang X, Crocker JT, Wang WL, Klibanski A
JournalJ Clin Endocrinol Metab
PubMed ID19383775
'CONTEXT: The mechanism of IGF2 overexpression in non-islet-cell tumor hypoglycemia is not understood. OBJECTIVE: We investigated the imprinting control and promoter usage for IGF2 expression to identify a mechanism for increased IGF-II production in non-islet-cell tumor hypoglycemia. PATIENT AND METHODS: A patient with metastatic hemangiopericytoma was studied. Tissue from the ... More
The role of interleukin-6 in lipopolysaccharide-induced fever by mechanisms independent of prostaglandin E2.
AuthorsNilsberth C, Elander L, Hamzic N, Norell M, Lonn J, Engstrom L, Blomqvist A
JournalEndocrinology
PubMed ID19022895
'Fever has been shown to be elicited by prostaglandin E(2) (PGE(2)) binding to its receptors on thermoregulatory neurons in the anterior hypothalamus. The signals that trigger PGE(2) production are thought to include proinflammatory cytokines, such as IL-6. However, although the presence of IL-6 is critical for fever, IL-6 by itself ... More
Host structural carbohydrate induces vector transmission of a bacterial plant pathogen.
AuthorsKilliny N, Almeida RP
JournalProc Natl Acad Sci U S A
PubMed ID20018775
'Many insect-borne pathogens have complex life histories because they must colonize both hosts and vectors for successful dissemination. In addition, the transition from host to vector environments may require changes in gene expression before the pathogen''s departure from the host. Xylella fastidiosa is a xylem-limited plant-pathogenic bacterium transmitted by leafhopper ... More
Inflammatory interaction between LIGHT and proteinase-activated receptor-2 in endothelial cells: potential role in atherogenesis.
AuthorsSandberg WJ, Halvorsen B, Yndestad A, Smith C, Otterdal K, Brosstad FR, Froland SS, Olofsson PS, Damas JK, Gullestad L, Hansson GK, Oie E, Aukrust P
JournalCirc Res
PubMed ID19023130
'The interaction between inflammatory cytokines and endothelial cells is a critical step in atherogenesis leading to endothelial dysfunction and inflammation. We have previously reported that the tumor necrosis factor superfamily member LIGHT could be involved in atherogenesis through its ability to promote vascular inflammation. In the present study we identified ... More
Elevated expression of osteopontin may be related to adipose tissue macrophage accumulation and liver steatosis in morbid obesity.
AuthorsBertola A, Deveaux V, Bonnafous S, Rousseau D, Anty R, Wakkach A, Dahman M, Tordjman J, Clement K, McQuaid SE, Frayn KN, Huet PM, Gugenheim J, Lotersztajn S, Le Marchand-Brustel Y, Tran A, Gual P
JournalDiabetes
PubMed ID18952835
'OBJECTIVE: Osteopontin (OPN) plays an important role in the development of insulin resistance and liver complications in dietary murine models. We aimed to determine the expression pattern of OPN and its receptor CD44 in obese patients and mice according to insulin resistance and liver steatosis. RESEARCH DESIGN AND METHODS: OPN ... More
Efflux-related resistance to norfloxacin, dyes, and biocides in bloodstream isolates of Staphylococcus aureus.
AuthorsDeMarco CE, Cushing LA, Frempong-Manso E, Seo SM, Jaravaza TA, Kaatz GW
JournalAntimicrob Agents Chemother
PubMed ID17576828
'Efflux is an important resistance mechanism in Staphylococcus aureus, but its frequency in patients with bacteremia is unknown. Nonreplicate bloodstream isolates were collected over an 8-month period, and MICs of four common efflux pump substrates, with and without the broad-spectrum efflux pump inhibitor reserpine, were determined (n = 232). A ... More
Bacillus polyfermenticus ameliorates colonic inflammation by promoting cytoprotective effects in colitic mice.
AuthorsIm E, Choi YJ, Pothoulakis C, Rhee SH
JournalJ Nutr
PubMed ID19675103
'Although human consumption of Bacillus polyfermenticus provides several health benefits, the probiotic effect of this bacterium against colonic inflammation has not yet, to our knowledge, been studied. Therefore, we induced colitis in mice by oral or intrarectal administration of dextran sodium sulfate (DSS) or trinitrobenzenosulfonic acid (TNBS), respectively, and investigated ... More
Exploration of tumor-suppressive microRNAs silenced by DNA hypermethylation in oral cancer.
AuthorsKozaki K, Imoto I, Mogi S, Omura K, Inazawa J
JournalCancer Res
PubMed ID18381414
'In the last few years, microRNAs (miRNA) have started a revolution in molecular biology and emerged as key players in the carcinogenesis. They have been identified in various tumor types, showing that different sets of miRNAs are usually deregulated in different cancers. To identify the miRNA signature that was specific ... More
Chronic furosemide or hydrochlorothiazide administration increases H+-ATPase B1 subunit abundance in rat kidney.
AuthorsNa KY, Kim GH, Joo KW, Lee JW, Jang HR, Oh YK, Jeon US, Chae SW, Knepper MA, Han JS
JournalAm J Physiol Renal Physiol
PubMed ID17311909
'Furosemide administration stimulates distal acidification. This has been attributed to the increased lumen-negative voltage in the distal nephron, but the aspect of regulatory mechanisms of H(+)-ATPase has not been clear. The purpose of this study is to investigate whether chronic administration of diuretics alters the expression of H(+)-ATPase and whether ... More
Targeting the cyclin E-Cdk-2 complex represses lung cancer growth by triggering anaphase catastrophe.
AuthorsGalimberti F, Thompson SL, Liu X, Li H, Memoli V, Green SR, DiRenzo J, Greninger P, Sharma SV, Settleman J, Compton DA, Dmitrovsky E
JournalClin Cancer Res
PubMed ID20028770
'PURPOSE: Cyclin-dependent kinases (Cdk) and their associated cyclins are targets for lung cancer therapy and chemoprevention given their frequent deregulation in lung carcinogenesis. This study uncovered previously unrecognized consequences of targeting the cyclin E-Cdk-2 complex in lung cancer. EXPERIMENTAL DESIGN: Cyclin E, Cdk-1, and Cdk-2 were individually ... More
Identification of transcriptional regulatory cascades in retinoic acid-induced growth arrest of HepG2 cells.
AuthorsNakanishi M, Tomaru Y, Miura H, Hayashizaki Y, Suzuki M
JournalNucleic Acids Res
PubMed ID18445634
'All-trans retinoic acid (ATRA) is a potent inducer of cell differentiation and growth arrest. Here, we investigated ATRA-induced regulatory cascades associated with growth arrest of the human hepatoma cell line HepG2. ATRA induced >2-fold changes in the expression of 402 genes including 55 linked to cell-cycle regulation, cell growth ... More
A novel correlation between LINE-1 hypomethylation and the malignancy of gastrointestinal stromal tumors.
AuthorsIgarashi S, Suzuki H, Niinuma T, Shimizu H, Nojima M, Iwaki H, Nobuoka T, Nishida T, Miyazaki Y, Takamaru H, Yamamoto E, Yamamoto H, Tokino T, Hasegawa T, Hirata K, Imai K, Toyota M, Shinomura Y
JournalClin Cancer Res
PubMed ID20978145
'PURPOSE: Gastrointestinal stromal tumors (GIST) are the most important mesenchymal tumors of the gastrointestinal tract. The vast majority of GISTs exhibit activating mutations of KIT or PDGFRA, but epigenetic alteration of GISTs is largely unknown. In this study, we aimed to clarify the involvement of DNA methylation in GIST malignancy. ... More
The transcription factor ATF4 promotes skeletal myofiber atrophy during fasting.
AuthorsEbert SM, Monteys AM, Fox DK, Bongers KS, Shields BE, Malmberg SE, Davidson BL, Suneja M, Adams CM
JournalMol Endocrinol
PubMed ID20197309
'Prolonged fasting alters skeletal muscle gene expression in a manner that promotes myofiber atrophy, but the underlying mechanisms are not fully understood. Here, we examined the potential role of activating transcription factor 4 (ATF4), a transcription factor with an evolutionarily ancient role in the cellular response to starvation. In mouse ... More
Molecular cloning and expression of the CRISP family of proteins in the boar.
AuthorsVadnais ML, Foster DN, Roberts KP
JournalBiol Reprod
PubMed ID18716287
'The family of mammalian cysteine-rich secretory proteins (CRISP) have been well characterized in the rat, mouse, and human. Here we report the molecular cloning and expression analysis of CRISP1, CRISP2, and CRISP3 in the boar. A partial sequence published in the National Center for Biotechnology Information (NCBI) database was ... More
Genome-wide identification of estrogen receptor alpha-binding sites in mouse liver.
AuthorsGao H, Falt S, Sandelin A, Gustafsson JA, Dahlman-Wright K
JournalMol Endocrinol
PubMed ID17901129
'We report the genome-wide identification of estrogen receptor alpha (ERalpha)-binding regions in mouse liver using a combination of chromatin immunoprecipitation and tiled microarrays that cover all nonrepetitive sequences in the mouse genome. This analysis identified 5568 ERalpha-binding regions. In agreement with what has previously been reported for human cell ... More
Blocking the Metabolism of Starch Breakdown Products in Arabidopsis Leaves Triggers Chloroplast Degradation
AuthorsStettler, M; Eicke, S; Mettler, T; Messerli, G; Hoertensteiner, S; Zeeman, SC
Journal
PubMed ID
'In most plants, a large fraction of photo-assimilated carbon is stored in the chloroplasts during the day as starch and remobilized during the subsequent night to support metabolism. Mutations blocking either starch synthesis or starch breakdown in Arabidopsis thaliana reduce plant growth. Maltose is the major product of starch breakdown ... More
A novel membrane fusion-mediated plant immunity against bacterial pathogens
AuthorsHatsugai, N; Iwasaki, S; Tamura, K; Kondo, M; Fuji, K; Ogasawara, K; Nishimura, M; Hara-Nishimura, I
Journal
PubMed ID
'Plants have developed their own defense strategies because they have no immune cells. A common plant defense strategy involves programmed cell death (PCD) at the infection site, but how the PCD-associated cell-autonomous immunity is executed in plants is not fully understood. Here we provide a novel mechanism underlying cell-autonomous immunity, ... More
Thrombospondin-1 peptide ABT-510 combined with valproic acid is an effective antiangiogenesis strategy in neuroblastoma.
AuthorsYang Q, Tian Y, Liu S, Zeine R, Chlenski A, Salwen HR, Henkin J, Cohn SL
JournalCancer Res
PubMed ID17308113
'In the pediatric cancer neuroblastoma, clinically aggressive disease is associated with increased levels of angiogenesis stimulators and high vascular index. We and others have hypothesized that blocking angiogenesis may be effective treatment for this pediatric malignancy. However, little is known about the efficacy of antiangiogenic agents in pediatric malignancies. Recently, ... More
IRAK-4 kinase activity is required for interleukin-1 (IL-1) receptor- and toll-like receptor 7-mediated signaling and gene expression.
AuthorsKoziczak-Holbro M, Joyce C, Gluck A, Kinzel B, Muller M, Tschopp C, Mathison JC, Davis CN, Gram H
JournalJ Biol Chem
PubMed ID17337443
'IRAK-4 is an essential component of the signal transduction complex downstream of the IL-1- and Toll-like receptors. Although regarded as the first kinase in the signaling cascade, the role of IRAK-4 kinase activity versus its scaffold function is still controversial. To investigate the role of IRAK-4 kinase function in vivo, ... More
Multiplex rt-PCR expression analysis of developmentally important genes in individual mouse preimplantation embryos and blastomeres.
AuthorsMay A, Kirchner R, Muller H, Hartmann P, El Hajj N, Tresch A, Zechner U, Mann W, Haaf T
JournalBiol Reprod
PubMed ID18784354
'We have developed a microfluidic chip-based qualitative assay for sensitive (10 RNA copies) detection of multiple transcripts in single cells. We determined the expression patterns of 17 developmentally important genes and isoforms in individual mouse preimplantation embryos from superovulated matings and blastomeres. The ubiquitously expressed histone variant H3f3a and ... More
Microarray analysis of developing flax hypocotyls identifies novel transcripts correlated with specific stages of phloem fibre differentiation.
AuthorsRoach MJ, Deyholos MK
JournalAnn Bot
PubMed ID18593690
'BACKGROUND AND AIMS: Hypocotyls are a commonly used model to study primary growth in plants, since post-germinative hypocotyls increase in size by cell elongation rather than cell division. Flax hypocotyls produce phloem fibres in bundles one to two cell layers thick, parallel to the protoxylem poles of the stele. ... More
Requirement of c-Jun NH2-Terminal Kinase for Ras-Initiated Tumor Formation.
AuthorsCellurale C, Sabio G, Kennedy NJ, Das M, Barlow M, Sandy P, Jacks T, Davis RJ
JournalMol Cell Biol
PubMed ID21282468
'The c-Jun NH(2)-terminal kinase (JNK) signal transduction pathway causes increased gene expression mediated, in part, by members of the activating transcription factor protein (AP1) group. JNK is therefore implicated in the regulation of cell growth and cancer. To test the role of JNK in Ras-induced tumor formation, we examined the ... More
Antiretroviral pre-exposure prophylaxis prevents vaginal transmission of HIV-1 in humanized BLT mice.
AuthorsDenton PW, Estes JD, Sun Z, Othieno FA, Wei BL, Wege AK, Powell DA, Payne D, Haase AT, Garcia JV
JournalPlos Med
PubMed ID18198941
'BACKGROUND: Worldwide, vaginal transmission now accounts for more than half of newly acquired HIV-1 infections. Despite the urgency to develop and implement novel approaches capable of preventing HIV transmission, this process has been hindered by the lack of adequate small animal models for preclinical efficacy and safety testing. Given the ... More
Target-dependent B7-H1 regulation contributes to clearance of central nervous system infection and dampens morbidity.
AuthorsPhares TW, Ramakrishna C, Parra GI, Epstein A, Chen L, Atkinson R, Stohlman SA, Bergmann CC
JournalJ Immunol
PubMed ID19380790
'The neurotropic coronavirus JHM strain of mouse hepatitis virus persists in oligodendroglia despite the presence of virus-specific CD8 T cells. Expression of programmed death 1 (PD-1) and B7-H1 were studied during acute and persistent infection to examine whether this negative regulatory mechanism contributes to CNS viral persistence. The majority of ... More
DOUBLETIME plays a noncatalytic role to mediate CLOCK phosphorylation and repress CLOCK-dependent transcription within the Drosophila circadian clock.
AuthorsYu W, Zheng H, Price JL, Hardin PE
JournalMol Cell Biol
PubMed ID19139270
'Circadian clocks keep time via gene expression feedback loops that are controlled by time-of-day-specific changes in the synthesis, activity, and degradation of transcription factors. Within the Drosophila melanogaster circadian clock, DOUBLETIME (DBT) kinase is necessary for the phosphorylation of PERIOD (PER), a transcriptional repressor, and CLOCK (CLK), a transcriptional activator, ... More
Growth factors of lower vertebrates: characterization of goldfish (Carassius auratus L.) macrophage colony-stimulating factor-1.
AuthorsHanington PC, Wang T, Secombes CJ, Belosevic M
JournalJ Biol Chem
PubMed ID17827160
'Colony-stimulating factor-1 (CSF-1) regulates mononuclear cell proliferation, differentiation, and survival. The functions of CSF-1 are well documented in mammals; however, little is known about CSF-1 biology in lower vertebrates. This is the first report on the identification and functional characterization of a fish CSF-1 molecule expressed highly in the ... More
Structure of the Plasmodium falciparum circumsporozoite protein, a leading malaria vaccine candidate.
AuthorsPlassmeyer ML, Reiter K, Shimp RL Jr, Kotova S, Smith PD, Hurt DE, House B, Zou X, Zhang Y, Hickman M, Uchime O, Herrera R, Nguyen V, Glen J, Lebowitz J, Jin AJ, Miller LH, MacDonald NJ, Wu Y, Narum DL
JournalJ Biol Chem
PubMed ID19633296
'The Plasmodium falciparum circumsporozoite protein (CSP) is critical for sporozoite function and invasion of hepatocytes. Given its critical nature, a phase III human CSP malaria vaccine trial is ongoing. The CSP is composed of three regions as follows: an N terminus that binds heparin sulfate proteoglycans, a four amino acid ... More
Overexpression of endoplasmic reticulum protein 29 regulates mesenchymal-epithelial transition and suppresses xenograft tumor growth of invasive breast cancer cells
AuthorsBambang, IF; Xu, SC; Zhou, JB; Salto-Tellez, M; Sethi, SK; Zhang, DH
Journal
PubMed ID
'Endoplasmic reticulum protein 29 (ERp29) is a novel endoplasmic reticulum ( ER) secretion factor that facilitates the transport of secretory proteins in the early secretory pathway. Recently, it was found to be overexpressed in several cancers; however, little is known regarding its function in breast cancer progression. In this study, ... More
B cell receptor-induced phosphorylation of Pyk2 and focal adhesion kinase involves integrins and the Rap GTPases and is required for B cell spreading.
AuthorsTse KW, Dang-Lawson M, Lee RL, Vong D, Bulic A, Buckbinder L, Gold MR
JournalJ Biol Chem
PubMed ID19561089
'Signaling by the B cell receptor (BCR) promotes integrin-mediated adhesion and cytoskeletal reorganization. This results in B cell spreading, which enhances the ability of B cells to bind antigens and become activated. Proline-rich tyrosine kinase (Pyk2) and focal adhesion kinase (FAK) are related cytoplasmic tyrosine kinases that regulate cell adhesion, ... More
miR-124 and miR-203 are epigenetically silenced tumor-suppressive microRNAs in hepatocellular carcinoma.
AuthorsFuruta M, Kozaki KI, Tanaka S, Arii S, Imoto I, Inazawa J
JournalCarcinogenesis
PubMed ID19843643
'MicroRNAs (miRNAs) are a class of small non-coding RNAs that, in general, negatively regulate gene expression. They have been identified in various tumor types, showing that different sets of miRNAs are usually deregulated in different cancers. Some miRNA genes harboring CpG islands undergo methylation-mediated silencing, a characteristic of many tumor ... More
Detection of Metastatic Head and Neck Squamous Cell Carcinoma Using the Relative Expression of Tissue-Specific Mir-205
AuthorsFletcher, AM; Heaford, AC; Trask, DK
Journal
PubMed ID
'The presence of cervical lymph node metastases in head and neck squamous cell carcinoma (HNSCC) is the strongest determinant of patient prognosis. Owing to the impact of nodal metastases on patient survival, a system for sensitive and accurate detection is required. Clinical staging of lymph nodes is far less accurate ... More
Purification of Nucleic Acids from Whole Blood Using Isotachophoresis
AuthorsPersat, A; Marshall, LA; Santiago, JG
Journal
PubMed ID
'We present and demonstrate a novel technique for the purification of nucleic acids from biological samples using isotachophoresis (ITP). We demonstrate a simple and rapid method to achieve ITP-based extraction, preconcentration, and purification of DNA from nanoliter volumes of whole blood. We show that ITP purification yields genomic DNA samples ... More
Trichothecene mycotoxins activate inflammatory response in human macrophages.
AuthorsKankkunen P, Rintahaka J, Aalto A, Leino M, Majuri ML, Alenius H, Wolff H, Matikainen S
JournalJ Immunol
PubMed ID19414795
'Damp building-related illnesses have caused concern for years in many countries. Although the problem is extensive, the knowledge of the immunological reactions behind damp building-related illnesses is still quite limited. Trichothecene mycotoxins form one major group of toxins, which possibly contribute to the illnesses. Stachybotrys chartarum is a well-known, ... More
Down-regulation of suppressor of cytokine signaling-3 causes prostate cancer cell death through activation of the extrinsic and intrinsic apoptosis pathways.
AuthorsPuhr M, Santer FR, Neuwirt H, Susani M, Nemeth JA, Hobisch A, Kenner L, Culig Z
JournalCancer Res
PubMed ID19738059
'Suppressor of cytokine signaling-3 (SOCS-3) acts as a negative feedback regulator of the Janus-activated kinase/signal transducers and activators of transcription factors signaling pathway and plays an important role in the development and progression of various cancers. To better understand the role of SOCS-3 in prostate cancer, SOCS-3 expression was ... More
MicroRNA-98 and let-7 confer cholangiocyte expression of cytokine-inducible Src homology 2-containing protein in response to microbial challenge.
AuthorsHu G, Zhou R, Liu J, Gong AY, Eischeid AN, Dittman JW, Chen XM
JournalJ Immunol
PubMed ID19592657
'Posttranscriptional gene regulation by microRNAs (miRNAs) has been implicated in the fine-tuning of TLR-mediated inflammatory response. The cytokine-inducible Src homology 2-containing protein (CIS), one member of the suppressors of cytokine signaling family of proteins, is an important negative regulator for inflammatory cytokine signaling. Using in vitro models using normal ... More
The sympathetic nervous system modulates CD4(+)FoxP3(+) regulatory T cells via a TGF-β-dependent mechanism
AuthorsBhowmick, S; Singh, A; Flavell, RA; Clark, RB; O'Rourke, J; Cone, RE
Journal
PubMed ID
'CD4(+)FoxP3(+) Tregs are essential mediators of the peripheral immune response to self-antigens. Accordingly, the homeostatic regulation of Treg activity and number would impact on the immune response to both self-and non-self antigens. Because the sympathetic nervous system (SNS) interacts chemically and physically with the central and peripheral immune system and ... More
Regulation of MRP2/ABCC2 and BSEP/ABCB11 expression in sandwich cultured human and rat hepatocytes exposed to inflammatory cytokines TNF-{alpha}, IL-6, and IL-1{beta}.
AuthorsDiao L, Li N, Brayman TG, Hotz KJ, Lai Y
JournalJ Biol Chem
PubMed ID20702406
'In the present study MRP2/ABCC2 and BSEP/ABCB11 expression were investigated in sandwich cultured (SC) human and rat hepatocytes exposed to the proinflammatory cytokines. The investigation was also done in lipopolysaccharide (LPS)-treated rats. In SC human hepatocytes, both absolute protein and mRNA levels of MRP2/ABCC2 were significantly down-regulated by TNF-alpha, IL-6, ... More
The capsid proteins of Aleutian mink disease virus activate caspases and are specifically cleaved during infection.
AuthorsCheng F, Chen AY, Best SM, Bloom ME, Pintel D, Qiu J
JournalJ Virol
PubMed ID20042496
'Aleutian mink disease virus (AMDV) is currently the only known member of the genus Amdovirus in the family Parvoviridae. It is the etiological agent of Aleutian disease of mink. We have previously shown that a small protein with a molecular mass of approximately 26 kDa was present during AMDV infection ... More
Time course of alterations in myocardial glucose utilization in the Zucker diabetic fatty rat with correlation to gene expression of glucose transporters: a small-animal PET investigation.
AuthorsShoghi KI, Gropler RJ, Sharp T, Herrero P, Fettig N, Su Y, Mitra MS, Kovacs A, Finck BN, Welch MJ
JournalJ Nucl Med
PubMed ID18632819
'Diabetic cardiomyopathy is associated with abnormalities in glucose metabolism. We evaluated myocardial glucose metabolism in a rodent model of type 2 diabetes, namely the Zucker diabetic fatty (ZDF) rat, and validated PET measurements of glucose uptake against gene and protein expression of glucose transporters (GLUTs). METHODS: Six lean and ... More
Effects of Increased Renal Tubular Vascular Endothelial Growth Factor (VEGF) on Fibrosis, Cyst Formation, and Glomerular Disease
AuthorsHakroush, S; Moeller, MJ; Theilig, F; Kaissling, B; Sijmonsma, TP; Jugold, M; Akeson, AL; Traykova-Brauch, M; Hosser, H; Haehnel, B; Groene, HJ; Koesters, R; Kriz, W
Journal
PubMed ID
'The role of vascular endothelial growth factor (VEGF) in renal fibrosis, tubular cyst formation, and glomerular diseases is incompletely understood. We studied a new conditional transgenic mouse system [Pax8-rtTA/(tetO)(7)VEGF], which allows increased tubular VEGF production in adult mice. The following pathology was observed. The interstitial changes consisted of a ubiquitous ... More
Immediate and sustained blood pressure lowering by urocortin 2: a novel approach to antihypertensive therapy?
AuthorsDieterle T, Meili-Butz S, Buhler K, Morandi C, John D, Buser PT, Rivier J, Vale WW, Peterson KL, Brink M
JournalHypertension
PubMed ID19204182
'Recently, novel corticotropin-releasing factor-related peptides, named urocortin 1, 2, and 3, and a distinct cardiac and peripheral vascular receptor (corticotropin-releasing factor receptor 2) were described being part of a peripheral corticotropin-releasing factor system modulating cardiovascular function in response to stress. Vasorelaxation and blood pressure lowering have been reported after ... More
Identification of DNA regions and a set of transcriptional regulatory factors involved in transcriptional regulation of several human liver-enriched transcription factor genes.
AuthorsMiura H, Tomaru Y, Nakanishi M, Kondo S, Hayashizaki Y, Suzuki M
JournalNucleic Acids Res
PubMed ID19074951
'Mammalian tissue- and/or time-specific transcription is primarily regulated in a combinatorial fashion through interactions between a specific set of transcriptional regulatory factors (TRFs) and their cognate cis-regulatory elements located in the regulatory regions. In exploring the DNA regions and TRFs involved in combinatorial transcriptional regulation, we noted that individual ... More
Intracellular cholesterol transporter StarD4 binds free cholesterol and increases cholesteryl ester formation.
AuthorsRodriguez-Agudo D, Ren S, Wong E, Marques D, Redford K, Gil G, Hylemon P, Pandak WM
JournalJ Lipid Res
PubMed ID18403318
'StarD4 protein is a member of the StarD4 subfamily of steroidogenic acute regulatory-related lipid transfer (START) domain proteins that includes StarD5 and StarD6, proteins whose functions remain poorly defined. The objective of this study was to isolate and characterize StarD4''s sterol binding and to determine in a hepatocyte culture model ... More
Rescue of progranulin deficiency associated with frontotemporal lobar degeneration by alkalizing reagents and inhibition of vacuolar ATPase.
AuthorsCapell A, Liebscher S, Fellerer K, Brouwers N, Willem M, Lammich S, Gijselinck I, Bittner T, Carlson AM, Sasse F, Kunze B, Steinmetz H, Jansen R, Dormann D, Sleegers K, Cruts M, Herms J, Van Broeckhoven C, Haass C
JournalJ Neurosci
PubMed ID21289198
'Numerous loss-of-function mutations in the progranulin (GRN) gene cause frontotemporal lobar degeneration with ubiquitin and TAR-DNA binding protein 43-positive inclusions by reduced production and secretion of GRN. Consistent with the observation that GRN has neurotrophic properties, pharmacological stimulation of GRN production is a promising approach to rescue GRN haploinsufficiency and ... More
ARABIDOPSIS TRITHORAX-RELATED7 Is Required for Methylation of Lysine 4 of Histone H3 and for Transcriptional Activation of FLOWERING LOCUS C
AuthorsTamada, Y; Yun, JY; Woo, SC; Amasino, RM
Journal
PubMed ID
'In the winter-annual accessions of Arabidopsis thaliana, presence of an active allele of FRIGIDA (FRI) elevates expression of FLOWERING LOCUS C (FLC), a repressor of flowering, and thus confers a vernalization requirement. FLC activation by FRI involves methylation of Lys 4 of histone H3 (H3K4) at FLC chromatin. Many multicellular ... More
Global identification of bursicon-regulated genes in Drosophila melanogaster.
AuthorsAn S, Wang S, Gilbert LI, Beerntsen B, Ellersieck M, Song Q
JournalBmc Genomics
PubMed ID18801173
'BACKGROUND: Bursicon is a heterodimer neuropeptide responsible for regulating cuticle sclerotization and wing expansion in several insect species. Recent studies indicate that the action of bursicon is mediated by a specific G protein-coupled receptor DLGR2 and the cAMP/PKA signaling pathway. However, little is known regarding the genes that are regulated ... More
Characterization of pharmacologically active compounds that inhibit poliovirus and enterovirus 71 infectivity.
AuthorsArita M, Wakita T, Shimizu H
JournalJ Gen Virol
PubMed ID18796721
'Poliovirus (PV) and enterovirus 71 (EV71) cause severe neurological symptoms in their infections of the central nervous system. To identify compounds with anti-PV and anti-EV71 activities that would not allow the emergence of resistant mutants, we performed drug screening by utilizing a pharmacologically active compound library targeting cellular factors ... More
Noninvasive imaging of cell proliferation following mitogenic extracellular kinase inhibition by PD0325901.
AuthorsLeyton J, Smith G, Lees M, Perumal M, Nguyen QD, Aigbirhio FI, Golovko O, He Q, Workman P, Aboagye EO
JournalMol Cancer Ther
PubMed ID18790789
'The mitogenic extracellular kinase 1/2 (MEK1/2) inhibitor, PD0325901, has potent activity in a number of cancer cell types in vitro. In SKMEL-28 human melanoma cells (BRAF mutant), the drug rapidly decreased phosphorylated extracellular signal-regulated kinase 1/2, cyclin D1, and thymidine kinase 1 protein levels. We investigated if 3''-deoxy-3''-[18F]fluorothymidine-positron emission ... More
15-hydroxyprostaglandin dehydrogenase is down-regulated in gastric cancer.
AuthorsThiel A, Ganesan A, Mrena J, Junnila S, Nykanen A, Hemmes A, Tai HH, Monni O, Kokkola A, Haglund C, Petrova TV, Ristimaki A
JournalClin Cancer Res
PubMed ID19584167
'PURPOSE: We have investigated the expression and regulation of 15-hydroxyprostaglandin dehydrogenase (15-PGDH) in gastric cancer. EXPERIMENTAL DESIGN: Clinical gastric adenocarcinoma samples were analyzed by immunohistochemistry and quantitative real-time PCR for protein and mRNA expression of 15-PGDH and for methylation status of 15-PGDH promoter. The effects of interleukin-1beta (IL-1beta) and epigenetic ... More
Resveratrol, an activator of SIRT1, upregulates sarcoplasmic calcium ATPase and improves cardiac function in diabetic cardiomyopathy.
AuthorsSulaiman M, Matta MJ, Sunderesan NR, Gupta MP, Periasamy M, Gupta M
JournalAm J Physiol Heart Circ Physiol
PubMed ID20008278
'Reduced sarcoplasmic calcium ATPase (SERCA2a) expression has been shown to play a significant role in the cardiac dysfunction in diabetic cardiomyopathy. The mechanism of SERCA2a repression is, however, not known. This study was designed to examine the effect of resveratrol (RSV), a potent activator of SIRT1, on cardiac function and ... More
Structural and functional analysis of the Entamoeba histolytica EhrabB gene promoter.
AuthorsRomero-Diaz M, Gomez C, Lopez-Reyes I, Martinez MB, Orozco E, Rodriguez MA
JournalBmc Mol Biol
PubMed ID17883848
'BACKGROUND: The Entamoeba histolytica EhrabB gene encodes for a Rab GTPase involved in phagocytosis. It is located at a virulence locus where the Ehcp112 gene is in the complementary strand at 332 bp of EhrabB start codon, suggesting a finely regulated transcription of both genes. However, the transcription regulation in ... More
Glut2-dependent glucose-sensing controls thermoregulation by enhancing the leptin sensitivity of NPY and POMC neurons.
AuthorsMounien L, Marty N, Tarussio D, Metref S, Genoux D, Preitner F, Foretz M, Thorens B
JournalFaseb J
PubMed ID20097878
'The physiological contribution of glucose in thermoregulation is not completely established nor whether this control may involve a regulation of the melanocortin pathway. Here, we assessed thermoregulation and leptin sensitivity of hypothalamic arcuate neurons in mice with inactivation of glucose transporter type 2 (Glut2)-dependent glucose sensing. Mice with inactivation of ... More
Using Cells Devoid of Ras Proteins as Tools for Drug Discovery
AuthorsUrosevic, J; Sum, EYM; Moneo, V; Drosten, M; Dhawahir, A; Becerra, M; Carnero, A; Barbacid, M
Journal
PubMed ID
'Mutational activation of RAS proteins occurs in nearly 30% of all human tumors. To date direct pharmacological inhibition of RAS oncoproteins has not been possible. As a consequence, current strategies are focusing on the development of inhibitors that target those kinases acting downstream of RAS proteins, including those of the ... More
BMP2-activated Erk/MAP kinase stabilizes Runx2 by increasing p300 levels and histone acetyltransferase activity.
AuthorsJun JH, Yoon WJ, Seo SB, Woo KM, Kim GS, Ryoo HM, Baek JH
JournalJ Biol Chem
PubMed ID20851880
'Runx2 is a critical transcription factor for osteoblast differentiation. Regulation of Runx2 expression levels and transcriptional activity is important for bone morphogenetic protein (BMP)-induced osteoblast differentiation. Previous studies have shown that extracellular signal-regulated kinase (Erk) activation enhances the transcriptional activity of Runx2 and that BMP-induced Runx2 acetylation increases Runx2 stability ... More
Borrelia burgdorferi lacking DbpBA exhibits an early survival defect during experimental infection.
AuthorsWeening EH, Parveen N, Trzeciakowski JP, Leong JM, Hook M, Skare JT
JournalInfect Immun
PubMed ID18809667
'Several Borrelia burgdorferi genes induced under mammalian host conditions have been purported to be important in Lyme disease pathogenesis based on their binding to host structures. These genes include the dbpBA locus, whose products bind host decorin and glycosoaminoglycans. Recently, the dbpBA genes were reported to be involved in borrelial ... More