GeneAmp™ 10X PCR Buffer I, (2 x 75 mL)
Applied Biosystems™

GeneAmp™ 10X PCR Buffer I, (2 x 75 mL)

Applied Biosystems GeneAmp 10X PCR Buffer I is optimized for use with AmpliTaq DNA Polymerase, resulting in robust PCR amplification.
Have Questions?
Catalog NumberQuantity
43798762 x 75 mL
Catalog number 4379876
Price (USD)
273.65
Online exclusive
291.00
Save 17.35
Each
Quantity:
2 x 75 mL
Request bulk or custom format
Price (USD)
273.65
Online exclusive
291.00
Save 17.35
Each
Applied Biosystems GeneAmp 10X PCR Buffer I is optimized for use with AmpliTaq DNA Polymerase, resulting in robust PCR amplification. GeneAmp 10X PCR Buffer I contains 15 mM MgCl2. When diluted 1:10, it provides the optimal pH and ionic strength for PCR amplification with AmpliTaq DNA Polymerase.

GeneAmp 10X PCR Buffer I is available in several convenient sizes and when used with AmpliTaq DNA Polymerase offers maximum PCR performance.

Note

See user's manual or package insert for limited label license and trademark information.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Fidelity (vs. Taq)1X
PolymeraseAmpliTaq DNA Polymerase
Product TypePCR Buffer
Quantity2 x 75 mL
Reaction FormatKit
Shipping ConditionWet Ice
Volume150 mL
Concentration10X
For Use With (Application)Standard PCR
Unit SizeEach
Contents & Storage
• GeneAmp 10X PCR Buffer I (with 15 mM MgCl2), 2 x 75 mL

Store at -15°C to -25°C.

Frequently asked questions (FAQs)

How many freeze cycles do you recommend for Applied Biosystems, GeneAmp 10X PCR Buffer I (Cat. No. 4379876)?

We do not have specific guidance on the number of freeze thaws. We recommend following the guidance for aliquoting 1.5 mL of this buffer into autoclaved 2 mL tubes after the first thaw as described in Product Insert: GeneAmp 10X PCR Buffer I. This will help increase efficiency when working with the product and serve to limit the number of freeze-thaws for the buffer.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.

What are the differences between GeneAmp 10X PCR Buffer I, PCR Buffer II and PCR Gold Buffer, and what are the formulations of each buffer?

The ionic strength and the pH of GeneAmp 10X PCR Gold Buffer have been carefully optimized for use with the AmpliTaq Gold enzyme. GeneAmp 10X PCR Buffer I contains Mg; Geneamp 10X PCR Buffer II and Geneamp 10X PCR Gold Buffer do not have Mg in them, but come with a separate vial of 25 mM MgCl2 so that you can titrate the Mg in your PCR reaction.

Buffer formulations:
Buffer I (500 mM KCl, 100 mM Tris-HCl, pH 8.3, 15 mM MgCl2, 0.01% (w/v) gelatin)
Buffer II (500 mM KCl, 100 mM Tris-HCl, pH 8.3), with separate tube of 25 mM MgCl2
Gold Buffer (500 mM KCl, 150 mM Tris-HCl pH 8.0), with separate tube of 25 mM MgCl2

Find additional tips, troubleshooting help, and resources within our PCR and cDNA Synthesis Support Center.