5-(chlorosulfonyl)-2-fluorobenzoic acid, 97%, Thermo Scientific Chemicals
5-(chlorosulfonyl)-2-fluorobenzoic acid, 97%, Thermo Scientific Chemicals
5-(chlorosulfonyl)-2-fluorobenzoic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-(chlorosulfonyl)-2-fluorobenzoic acid, 97%, Thermo Scientific Chemicals

CAS: 37098-75-2 | C7H4ClFO4S | 238.613 g/mol
Have Questions?
Catalog NumberQuantityPackaging
4380100505 gGlass Bottle
Catalog number 438010050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS37098-75-2
IUPAC Name5-(chlorosulfonyl)-2-fluorobenzoic acid
Molecular FormulaC7H4ClFO4S
InChI KeyCISMZCVQNVNWJW-UHFFFAOYSA-N
SMILESOC(=O)C1=C(F)C=CC(=C1)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Elemental Analysis1.62 to 1.76 % H
Elemental Analysis33.82 to 36.64 % C
Appearance (Form)Crystalline powder
Appearance (Color)White to off-white
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only