Do you have stability information for the FAST MicroSEQ D2 rDNA Fungal PCR Kit (Cat. No. 4382397) when subjected to temperature fluctuations?
The FAST MicroSEQ D2 rDNA Fungal PCR Kit is stable if stored at -20 degrees C for 15-16 months. We do not have any information available for temperatures outside of this range.