tert-Butyl 4-(2-chloroethyl)piperazine-1-carboxylate, 97%
tert-Butyl 4-(2-chloroethyl)piperazine-1-carboxylate, 97%
tert-Butyl 4-(2-chloroethyl)piperazine-1-carboxylate, 97%
Thermo Scientific Chemicals

tert-Butyl 4-(2-chloroethyl)piperazine-1-carboxylate, 97%

CAS: 208167-83-3 | C11H21ClN2O2 | 248.76 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 438240010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS208167-83-3
IUPAC Nametert-butyl 4-(2-chloroethyl)piperazine-1-carboxylate
Molecular FormulaC11H21ClN2O2
InChI KeyMYOWELLYEZMECA-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)N1CCN(CCCl)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light brown
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Melting point62°C to 68°C
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only