You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
New Products
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
Home
▸
Shop All Products
▸
Custom Products and Services
▸
Custom RNA Products
▸
Carrier RNA
Applied Biosystems™
Carrier RNA
Ambion® Carrier RNA (1 mg/mL) is a component of all the MagMAX™ Isolation Kits that may be purchased separately.
Provided
Read more
Have Questions?
Catalog Number
Quantity
4382878
500 μL
Catalog number
4382878
Price (USD)
303.65
Online exclusive
337.00
Save 33.35
Each
Add to cart
Quantity:
500 μL
Price (USD)
303.65
Online exclusive
337.00
Save 33.35
Each
Add to cart
Ambion® Carrier RNA (1 mg/mL) is a component of all the MagMAX™ Isolation Kits that may be purchased separately. Provided in one tube of 500 μL.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)
Carrier for Nucleic Acid Precipitation
Product Line
Ambion
Product Type
Carrier RNA
Quantity
500 μL
Shipping Condition
Dry Ice
Unit Size
Each
Contents & Storage
Store solution at –20°C.
Frequently asked questions (FAQs)
I ran out of carrier RNA for my MagMAX isolation kit. Can I order this part separately?
Yes, carrier RNA can be ordered separately. The Cat. No. is 4382878.