BioProbes™ Journal of Cell Biology Applications—Subscribe today »
Get upto 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits
Streptavidin-CY3 (ZyMax Grade)
Invitrogen™

Streptavidin-CY3 (ZyMax Grade)

Streptavidin-CY3 (ZyMax Grade)
Have Questions?
Catalog NumberQuantity
438315
also known as 43-8315
1 mL
Catalog number 438315
also known as 43-8315
Price (USD)
458.00
Each
Quantity:
1 mL
Price (USD)
458.00
Each

Applications: Immunohistochemistry, Western blotting, ELISA, ISH

ZyMAX™ reagents offer enhanced assay sensitivity, higher titer, and greater flexibility in assay design than standard grade reagents.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Concentration1 mg/mL
Product TypeStreptavidin Conjugate (fluorescent)
Quantity1 mL
Shipping ConditionWet Ice
ConjugateCy3
FormLiquid
Product LineInvitrogen
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C).