Product Image
Applied Biosystems™

HP Deskjet Printer Model 6940 (100-240 V)

This is a Hewlett-Packard Deskjet Printer model 6940 (100-240V) for Genetic Analyzers.
Have Questions?
Catalog NumberQuantity
43836741 Ea.
Catalog number 4383674
Price (USD)
374.00
Each
Quantity:
1 Ea.
Price (USD)
374.00
Each

This is a Hewlett-Packard Deskjet Printer model 6940 (100-240V) for Genetic Analyzers.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
For Use With (Application)Sequencing
For Use With (Equipment)310 Genetic Analyzer, 3100 Genetic Analyzer
Product TypeHP Deskjet Printer Model
Quantity1 Ea.
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
Store at room temperature.