TAQMAN PREAMP MASTER MIX (2X) - FAQs
TAQMAN PREAMP MASTER MIX (2X) - FAQs
View additional product information for TAQMAN PREAMP MASTER MIX (2X) - FAQs (4384266)
1 product FAQs found
How do I reorder the TaqMan PreAmp Master Mix, Cat No. 4384266? It does not match the Cat. No. when I search online.
This is an internal part number for the TaqMan PreAmp Master Mix. To order, please use Cat No. 4391128 for the 40 reaction size or Cat. No. 4488593 for the 200 reaction size. This is the same product.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.