3-Methyl-3-penten-2-one, 98%
3-Methyl-3-penten-2-one, 98%
3-Methyl-3-penten-2-one, 98%
Thermo Scientific Chemicals

3-Methyl-3-penten-2-one, 98%

CAS: 565-62-8 | C6H10O | 98.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 438440100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS565-62-8
IUPAC Name(3Z)-3-methylpent-3-en-2-one
Molecular FormulaC6H10O
InChI KeyZAMCMCQRTZKGDX-PLNGDYQASA-N
SMILESC\C=C(\C)C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only