Strontium nitrate, 98%, extra pure
Strontium nitrate, 98%, extra pure
Strontium nitrate, 98%, extra pure
Thermo Scientific Chemicals

Strontium nitrate, 98%, extra pure

CAS: 10042-76-9 | N2O6Sr | 211.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Packaging:
Plastic bottle
Catalog number 438520010
Price (USD)
-
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS10042-76-9
IUPAC Namestrontium(2+) dinitrate
Molecular FormulaN2O6Sr
InChI KeyDHEQXMRUPNDRPG-UHFFFAOYSA-N
SMILES[Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Complexometric>=97.5 %
Loss on drying=<0.5 %
Heavy metals (as Pb)=<10 ppm
pH5 to 7 (5 % at 20°C)
Appearance (Form)Crystalline powder or crystals
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only