Fast SYBR™ Green Master Mix, 1 x 5 mL - FAQs
Fast SYBR™ Green Master Mix, 1 x 5 mL - FAQs
View additional product information for Fast SYBR™ Green Master Mix - FAQs (4385610, 4385612, 4385614, 4385616, 4385618, 4385617)
2 product FAQs found
Is there a SYBR Green Fast Master Mix currently available?
Yes. The following reagents can be used for fast cycling: Fast SYBR Green Master Mix, EXPRESS SYBR GreenER qPCR Supermix Universal, and SYBR GreenER qPCR SuperMix for ABI PRISM instruments.
What is the synthesis rate of AmpliTaq Gold Fast DNA Polymerase LD included in the Fast SYBR Green Master Mix (Cat. No. 4385612)?
DNA synthesis rate of the AmpliTaq Gold Fast DNA Polymerase LD is 1 min/kb.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.