Nunc™ MicroWell™ MiniTrays
Nunc™ MicroWell™ MiniTrays
Nunc™ MicroWell™ MiniTrays
Nunc™ MicroWell™ MiniTrays
Nunc™ MicroWell™ MiniTrays
Nunc™ MicroWell™ MiniTrays
Thermo Scientific™

Nunc™ MicroWell™ MiniTrays

Create more space in lab freezers and inubators with Nunc MicroWell MiniTrays, the optical clarity of which allows the trays to be used with inverted microscopes.
Catalog NumberWell CountClosure IncludedSterility
43873372YesNon-sterile
43922560YesNon-sterile
45225660YesSterile
47037872YesNon-sterile
47240060NoSterile
Catalog number 438733
Price (JPY)
26,340
Case of 100
Well Count:
72
Closure Included:
Yes
Sterility:
Non-sterile

Create more space in lab freezers and inubators with Thermo Scientific™ Nunc™ MicroWell™ MiniTrays, the optical clarity of which allows the trays to be used with inverted microscopes.

  • Limited adsorption of proteins
  • Lids have keyed tabs for proper orientation
  • Large raised alphanumeric coordinates
  • Unique surface treatment to maximize sample and reagent mixing
  • MiniTrays for serological applications have slightly smaller well dimensions
Specifications
Closure IncludedYes
Closure MaterialPolystyrene
For Use With (Application)For serological applications
IncludesLid
Length (Metric)84 mm
No. per Case100
No. per Pack10
ShapeRectangular
TypeCell Culture Dish
Width (Metric)59 mm
Bottom ShapeFlat
Height (Metric)59 mm
MaterialPolystyrene
SterilityNon-sterile
Surface TreatmentCell Culture
Well Count72
Unit SizeCase of 100