2-Methyl-2-phenylpropylmagnesium chloride, 0.5M solution in diethyl ether, AcroSeal™
2-Methyl-2-phenylpropylmagnesium chloride, 0.5M solution in diethyl ether, AcroSeal™
2-Methyl-2-phenylpropylmagnesium chloride, 0.5M solution in diethyl ether, AcroSeal™
2-Methyl-2-phenylpropylmagnesium chloride, 0.5M solution in diethyl ether, AcroSeal™
Thermo Scientific Chemicals

2-Methyl-2-phenylpropylmagnesium chloride, 0.5M solution in diethyl ether, AcroSeal™

CAS: 35293-35-7 | C10H13ClMg | 192.97 g/mol
Have Questions?
Catalog NumberQuantityPackaging
438751000100 mLAcroSeal™ Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 438751000
Price (USD)
-
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS35293-35-7
IUPAC Namechloro(2-methyl-2-phenylpropyl)magnesium
Molecular FormulaC10H13ClMg
InChI KeyBXKYFONHSSFLKR-UHFFFAOYSA-M
SMILESCC(C)(C[Mg]Cl)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Appearanceatmosphere
Appearance (Color)Colorless to yellow to grey
AppearanceIf precipitation is present, gently mix
AppearanceMay develop some turbidity or precipitate
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only