DEPC-Treated Water, 4 x 1000 mL - FAQs
DEPC-Treated Water, 4 x 1000 mL - FAQs
View additional product information for DEPC-Treated Water - FAQs (AM9906, AM9916, 4387937, AM9920, AM9922, AM9915G)
1 product FAQs found
How can DEPC-treated water be prepared?
DEPC-treated (ribonuclease free) water can be prepared as decribed below:
Add 1 mL DEPC per L of H2O
Shake until DEPC has dissolved in the water
Let sit for 1 h
Autoclave
Buffers containing Tris cannot be DEPC-treated (Tris inactivates DEPC by binding to amino groups). To prepare a Tris buffer, the water should be DEPC-treated and autoclaved before adding Tris. After addition of the Tris, the solution should be autoclaved again.