1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide, 97%
1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide, 97%
1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide, 97%
Thermo Scientific Chemicals

1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide, 97%

CAS: 1892-57-5 | C8H17N3 | 155.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Catalog number 438840100
Price (USD)
207.65
Online exclusive
230.00
Save 22.35
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
207.65
Online exclusive
230.00
Save 22.35
Each
Chemical Identifiers
CAS1892-57-5
IUPAC Name({[3-(dimethylamino)propyl]imino}methylidene)(ethyl)amine
Molecular FormulaC8H17N3
InChI KeyLMDZBCPBFSXMTL-UHFFFAOYSA-N
SMILESCCN=C=NCCCN(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4590 to 1.4630 (20°C, 589 nm)
GC>=96.0 %
Appearance (Color)Colorless to light yellow
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only