High Capacity RNA-to-cDNA Master Mix - FAQs
High Capacity RNA-to-cDNA Master Mix - FAQs
View additional product information for High Capacity RNA-to-cDNA Master Mix - FAQs (4390777)
1 product FAQs found
Can cDNA be generated using the High Capacity cDNA Reverse Transcription kit (SKU 4322171) for quantitative qPCR applications?
Yes, the single-stranded cDNA generated via the High-Capacity cDNA Reverse Transcription can be used in real-time PCR applications. In addition, we also recommend Superscript VILO cDNA Synthesis Kit, High Capacity RNA to cDNA kit, and/or High Capacity RNA to cDNA Master Mix for total RNA.