Anti-miR™ hsa-let-7c miRNA Inhibitor Positive Control
Ambion™ Anti-miR™ miRNA Inhibitors are chemically modified, single-stranded nucleic acids designed to specifically bind to and inhibit endogenous microRNA (miRNA)자세히 알아보기
Have Questions?
카탈로그 번호
수량
4392431
5 nmol
카탈로그 번호 4392431
제품 가격(KRW)
534,000
온라인 행사
Ends: 30-Sep-2026
593,000
할인액 59,000 (10%)
Each
수량:
5 nmol
제품 가격(KRW)
534,000
온라인 행사
Ends: 30-Sep-2026
593,000
할인액 59,000 (10%)
Each
Ambion™ Anti-miR™ miRNA Inhibitors are chemically modified, single-stranded nucleic acids designed to specifically bind to and inhibit endogenous microRNA (miRNA) molecules. These ready-to-use inhibitors can be introduced into cells using transfection or electroporation parameters similar to those used for siRNAs. Anti-miR™ let-7c miRNA Inhibitor Positive Control provides a convenient, validated positive control for experiments using Anti-miR miRNA Inhibitors. Endogenous let-7 miRNA negatively regulates HMGA2, a ubiquitously expressed, nonhistone chromatin protein that modulates gene expression through changes in chromatin architecture. When transfected into human and mouse cells, Anti-miR hsa-let-7c miRNA Inhibitor blocks endogenous let-7c miRNA, resulting in increased levels of HMGA2 mRNA.
Accessory Products: Also available are Anti-miR™ miRNA Inhibitors (SKU #AM17000 and #AM17001). To select your miRNA sequence and order, please visit the GeneAssist™ miRNA Workflow Builder. Anti-miR™ miRNA Inhibitor Negative Control #1 (SKU #AM17010) is a random sequence that has been validated to produce no identifiable effects on known miRNA function. Cy™3 and FAM™ dye-labeled Anti-miR Negative Controls (SKU #AM17011 and #AM17012, respectively) are also available. The Pre-miR™ miRNA Starter Kit (SKU #AM1540) can be used to optimize transfection of Pre-miR™ miRNA Precursors.
For Research Use Only. Not for use in diagnostic procedures.
사양
용도(애플리케이션)miRNA & Non-Coding RNA Analysis
라벨 또는 염료Unconjugated
제품라인Ambion, Anti-miR
제품 유형miRNA Inhibitor
수량5 nmol
배송 조건Room Temperature
제어 유형Positive Control
RNAi TypemiRNA Inhibitor
Unit SizeEach
구성 및 보관
Anti-miR™ miRNA is provided dried down in a single tube, along with Nuclease-free Water for resuspension, and should be stored at –20°C. It is guaranteed for 1 year when stored dried at –20°C.