POP-7™ Polymer Dx
Applied Biosystems™
POP-7™ Polymer Dx
POP-7 Polymer Dx is pre-formulated in a single-use pouch.
Have Questions?
Change viewbuttonViewtableView
Quantity:
960 Samples
384 Samples
Catalog number 4393713
Price (USD)
864.65
Online exclusive
940.00
Save 75.35
Each
Quantity:
960 Samples
Price (USD)
864.65
Online exclusive
940.00
Save 75.35
Each
POP-7 Polymer Dx and POP-6 Polymer Dx are separation matrices for the Applied Biosystems Dx genetic analyzers. They are conveniently offered in easy-to-use pouch packages with either 960 samples (maximum) or 384 samples (maximum) size formats. Each polymer is optimized for specific applications.
Easy to use
The polymer is pre-formulated in a single use pouch.
Radio frequency identification label
A radio frequency identification (RFID) encodes easy-to-track information such as part and lot number, samples remaining, and expiry dates (displayed on software dashboard).
Robust formulation
POP-7 Polymer Dx and POP-6 Polymer Dx are formulated for a wide range of applications.
Reproducible results
POP-7 Polymer Dx and POP-6 polymers dynamically coat the capillary wall to control electro-osmotic flow. Their definite quality and uniform consistency eliminate guesswork and help ensure reproducibility. POP-7 Polymer Dx and POP-6 Polymer Dx are specifically formulated to separate DNA fragments of a known size range at a desired resolution and run time.
Many choices
POP-7 Polymer Dx is very versatile and designed for a wide range of short-read to long-read sequencing applications. POP-6 polymer is the choice for certain sequencing applications.
Performance optimized polymer
This product is intended for in-vitro diagnostic use on the Applied Biosystems Dx genetic analyzers and is not validated for use with other systems or applications. The Applied Biosystems Dx genetic analyzers are to be used by trained operators in the analysis of human DNA for the detection of fluorescently labeled DNA nucleotides by capillary electrophoresis. Refer to the appropriate Applied Biosystems Dx genetic analyzer user guide for further details.
For in-vitro diagnostic use only
Specifications
For Use With (Application)Long-Read Sequencing, Standard Sequencing, Short Read Sequencing, Fragment Analysis (Denaturing)
Product LinePOP-7
Recommended StorageStore the polymer pouch at 2-8°C until ready to use. Note that expired pouches cannot be used on the instrument. On-instrument lifeA/UsageB On 3500 Dx (8 capillary) Genetic Analyzer, -Lower of 7 days or 960 samples or 120 Injections On 3500xl Dx (24 capillary) Genetic Analyzer -Lower of 7 days or 960 samples or 50 Injections The pouch includes additional polymer volume to accommodate both installation and a reasonable number of post-installation wizard operations such as bubble removal without affecting the number of samples/injections available.
Quantity960 Samples
Unit SizeEach
Frequently asked questions (FAQs)
When I started my run on the Applied Biosystems 3500/3500xL Genetic Analyzer, an error occurred: "Unstable Electrophoresis current detected, check for air bubbles" but I don't see any bubbles. What else can cause this?
Some of the other causes of an Unstable Electrophoresis current detected error message are:
1.Leak on the system
2. Polymer that:
-Has expired
-Has been left on the instrument for more than the recommended time
-Is a mixture of expired polymer and non-expired polymer
3. Running Buffer that was:
-On the instrument longer than 2 weeks
-Not filled to the fill line or evaporated below the fill line
4. An arcing event that was not cleaned afterwards using the water wash wizard
5. Not performing regular maintenance on the instrument
6. Hardware issues
Inspect the system for leaks. If you do not see any leaks on the system, perform the wash the pump chamber and channels wizard using the conditioning pouch and place fresh, non-expired polymer and fresh Anode and Cathode buffer containers. If the problem persists, a service call may be required.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.
Do I need to replace the buffer every 14 days if the Applied Biosystems 3500/3500xL Genetic Analyzer is not in use?
If the instrument is not in use, it is not necessary to replace the buffer every 14 days. However, the buffer level needs to remain at the fill line to prevent the capillary or array from drying out. Top off the buffer volume with water if necessary. Replace the anode and cathode buffer containers prior to starting a new run.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.