L-3,4-Dihydroxyphenylalanine methyl ester hydrochloride, 95%
L-3,4-Dihydroxyphenylalanine methyl ester hydrochloride, 95%
L-3,4-Dihydroxyphenylalanine methyl ester hydrochloride, 95%
Thermo Scientific Chemicals

L-3,4-Dihydroxyphenylalanine methyl ester hydrochloride, 95%

CAS: 1421-65-4 | C10H14ClNO4 | 247.68 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 439490010
Price (USD)
153.65
Online exclusive
170.00
Save 16.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
153.65
Online exclusive
170.00
Save 16.35
Each
Chemical Identifiers
CAS1421-65-4
IUPAC Namehydrogen methyl 2-amino-3-(3,4-dihydroxyphenyl)propanoate chloride
Molecular FormulaC10H14ClNO4
InChI KeyWFGNJLMSYIJWII-UHFFFAOYNA-N
SMILES[H+].[Cl-].COC(=O)C(N)CC1=CC=C(O)C(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Elemental Analysis45.58 to 51.40 % (C) On dry basis
Elemental Analysis5.32 to 5.99 % (N) On dry basis
Water=<5.00 %
Appearance (Color)White to light brown
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only