4-Bromophenethyl bromide, 96%
4-Bromophenethyl bromide, 96%
4-Bromophenethyl bromide, 96%
Thermo Scientific Chemicals

4-Bromophenethyl bromide, 96%

CAS: 1746-28-7 | C8H8Br2 | 263.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
Catalog number 439530100
Price (USD)
-
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS1746-28-7
IUPAC Name1-bromo-4-(2-bromoethyl)benzene
Molecular FormulaC8H8Br2
InChI KeyAPTDRDYSJZQPPI-UHFFFAOYSA-N
SMILESBrCCC1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=95.0 %
Appearance (Form)Liquid or low melting solid
Appearance (Color)Colorless to yellow to light brown
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only