(1R,2R,3R,5S)-(-)-Isopinocampheylamine, 95%
(1R,2R,3R,5S)-(-)-Isopinocampheylamine, 95%
(1R,2R,3R,5S)-(-)-Isopinocampheylamine, 95%
Thermo Scientific Chemicals

(1R,2R,3R,5S)-(-)-Isopinocampheylamine, 95%

CAS: 69460-11-3 | C10H19N | 153.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 439540010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS69460-11-3
IUPAC Name2,6,6-trimethylbicyclo[3.1.1]heptan-3-amine
Molecular FormulaC10H19N
InChI KeyVPTSZLVPZCTAHZ-UHFFFAOYNA-N
SMILESCC1C2CC(CC1N)C2(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=94.0 %
Specific optical rotation-30.9° to -36.9° (20°C, 589 nm) (c=1, Chloroform)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only