Benzothiazole-6-carboxylic acid, 96%
Benzothiazole-6-carboxylic acid, 96%
Benzothiazole-6-carboxylic acid, 96%
Thermo Scientific Chemicals

Benzothiazole-6-carboxylic acid, 96%

CAS: 3622-35-3 | C8H5NO2S | 179.19 g/mol
Have Questions?
Catalog NumberQuantity
4397000505 g
Catalog number 439700050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS3622-35-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light brown
Appearance (Form)Powder
Infrared spectrumConforms
Melting point>=240.0°C (Dec)
HPLC>=95.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only