Product Image
Applied Biosystems™
MicroSEQ™ ID 16S rDNA 500 Library v2013
This separately-available MicroSEQ™ ID 16S rDNA 500 Library v2013 is used to update older versions of the library co-installed with MicroSEQ™ ID Microbial Identification Software.
Have Questions?
Catalog NumberQuantity
43982201 Kit
Catalog number 4398220
Price (USD)
18,680.00
Each
Quantity:
1 Kit
Price (USD)
18,680.00
Each
This separately-available MicroSEQ™ ID 16S rDNA 500 Library v2013 is used to update older versions of the library co-installed with MicroSEQ™ ID Microbial Identification Software. Purchase of this library is not required when purchasing the software, as the most recent library is included with most software purchases. The library contains 2100 reference 16S rRNA sequences from bacteria against which an unknown bacterial sequence can be compared using MicroSEQ™ ID analysis software in order to identify the unknown organism. The entries contain the first 500 bases of the 16S rRNA gene, and the library is regularly updated with new entries and current taxonomy. The first 500 bases are generally sufficient to identify bacterial unknowns.
Related Links:
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Data EntrySequencing run
Data OutputsBacterial Identification
For Use With (Application)Sequencing
For Use With (Equipment)3130xl Genetic Analyzer, 3730xl DNA Analyzer, 3130 Genetic Analyzer, 3730 DNA Analyzer, 3500xL Genetic Analyzer, 3500 Genetic Analyzer
LicenseSeat
Operating SystemWindows Vista, Windows XP (Service Pack 3), Windows 7
Product LineMicroSEQ
Product TyperDNA 500 Library
Quantity1 Kit
SoftwareSecondary Analysis
Unit SizeEach
Frequently asked questions (FAQs)
What are the differences in library entry format between MicroSeq ID 16S rDNA 500 Supplemental Library and MicroSEQ ID 16S rDNA 500 Library v2013?
The library entry format for MicroSeq ID 16S rDNA 500 Supplemental Library contains strain information in the Comments column. In addition, the Source column has NCBI information for all entries and the Entry Number column includes accession number for the sequences.
What is the MicroSeq ID 16S rDNA 500 Supplemental Library?
This library is an add-on to MicroSEQ ID 16S rDNA 500 Library v2013 (Cat. No. 4398220).
What is the recommended analysis when using MicroSeq ID 16S rDNA 500 Supplemental Library?
We recommend using MicroSeq ID 16S rDNA 500 Supplemental Library only when the MicroSEQ ID 16S rdna 500 Library v2013 database generates a match <99% but has met all other acceptance criteria.