2-Bromo-1-ethyl-pyridinium tetrafluoroborate, 97%
2-Bromo-1-ethyl-pyridinium tetrafluoroborate, 97%
2-Bromo-1-ethyl-pyridinium tetrafluoroborate, 97%
Thermo Scientific Chemicals

2-Bromo-1-ethyl-pyridinium tetrafluoroborate, 97%

CAS: 878-23-9 | C7H9BBrF4N | 273.86 g/mol
Have Questions?
Catalog NumberQuantity
4399600505 g
Catalog number 439960050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS878-23-9
IUPAC Name2-bromo-1-ethylpyridin-1-ium; tetrafluoroboranuide
Molecular FormulaC7H9BBrF4N
InChI KeyYJDXVQLBIAJTHP-UHFFFAOYSA-N
SMILESF[B-](F)(F)F.CC[N+]1=CC=CC=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point100°C to 108°C
HPLC>=96.0 %
Appearance (Color)White
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only