Nunc MaxiSorp™ flat-bottom - FAQs
Nunc MaxiSorp™ flat-bottom - FAQs
View additional product information for Nunc MaxiSorp™ flat-bottom - FAQs (44-2404-21)
1 product FAQs found
Is there an alternative to Nunc MaxiSorp flat-bottom 96 well plates (Cat. No. 44-2404-21)?
We recommend Thermo Scientific Clear Flat-Bottom Immuno Nonsterile 96-Well Plates (Cat. No. 442404) as an alternative to the discontinued Nunc Nunc MaxiSorp flat-bottom 96 well plates.