DNA Extract All Reagents Kit, 200 mL - FAQs
DNA Extract All Reagents Kit, 200 mL - FAQs
View additional product information for DNA Extract All Reagents Kit - FAQs (4402616, 4403319, 4402599)
1 product FAQs found