Precision Plate Holder for 0.1 mL Tube Strips (for 7500 Fast System) - FAQs
Precision Plate Holder for 0.1 mL Tube Strips (for 7500 Fast System) - FAQs
View additional product information for Precision Plate Holder for 0.1 mL Tube Strips (for 7500 Fast System) - FAQs (4403809)
1 product FAQs found
Can I use MicroAmp Optical Tubes on the Applied Biosystems 7500 Fast System?
Unfortunately, there are no single-tube formats currently available that will fit the Applied Biosystems 7500 Fast Real-Time PCR System thermal cycler block. Regular MicroAmp Optical Tubes (Cat. No. N801-0933) will not fit in the block and will get crushed. If you want to use tubes in the 7500F, please use the MicroAmp Fast 8-Tube Strip (Cat. No. 4358293) along with the Precision Plate Holder for Tubes (Cat. No. 4403809).
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.