1-Methyl-4-piperidinemethanol, 97%
1-Methyl-4-piperidinemethanol, 97%
1-Methyl-4-piperidinemethanol, 97%
Thermo Scientific Chemicals

1-Methyl-4-piperidinemethanol, 97%

CAS: 20691-89-8 | C7H15NO | 129.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 440480010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS20691-89-8
IUPAC Name(1-methylpiperidin-4-yl)methanol
Molecular FormulaC7H15NO
InChI KeyKJZLJGZZDNGGCA-UHFFFAOYSA-N
SMILESCN1CCC(CO)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Titration with HCl>=96.0 to =<104.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only