4,5-Dimethoxy-2-nitrobenzyl bromide, 97%
4,5-Dimethoxy-2-nitrobenzyl bromide, 97%
4,5-Dimethoxy-2-nitrobenzyl bromide, 97%
4,5-Dimethoxy-2-nitrobenzyl bromide, 97%
Thermo Scientific Chemicals

4,5-Dimethoxy-2-nitrobenzyl bromide, 97%

CAS: 53413-67-5 | C9H10BrNO4 | 276.09 g/mol
製品番号(カタログ番号) 440770050
価格(JPY)
-
見積もりを依頼する
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS53413-67-5
IUPAC Name1-(bromomethyl)-4,5-dimethoxy-2-nitrobenzene
Molecular FormulaC9H10BrNO4
InChI KeyUEKFEYNZISYRRH-UHFFFAOYSA-N
SMILESCOC1=CC(CBr)=C(C=C1OC)[N+]([O-])=O
さらに表示
Infrared spectrumConforms
HPLC>=96.0 %
Appearance (Form)Crystalline powder or needles
Appearance (Color)Yellow to brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only