My order of Cathode Buffer Container (Cat. No. 4408256) was shipped at room temperature, but the recommended storage conditions are 2-6 degrees C. Can I still use it?
Yes, it is still usable. We ship these buffers at room temperature to minimize the environmental impact of packaging and shipping these products on gel packs. We performed stability and performance studies and found that there was no impact to the performance of these buffers when shipped at room temperature. Please see the CE Buffers Ambient Shipping white paper detailing these studies, for your reference.