Cathode Buffer Container Dx
Applied Biosystems™

Cathode Buffer Container Dx

The Cathode Buffer Container Dx contains 1X running buffer to support electrophoresis applications on Applied Biosystems Dx genetic analyzers.
Have Questions?
Catalog NumberQuantity
44082584 Ea.
Catalog number 4408258
Price (USD)
313.00
Each
Quantity:
4 Ea.
Price (USD)
313.00
Each

The Cathode Buffer Container Dx is for use with Applied Biosystems Dx genetic analyzers.

The Cathode Buffer Container Dx contains 1X running buffer to support electrophoresis applications on Applied Biosystems Dx genetic analyzers. The Cathode Buffer Container Dx has two separate compartments: the left side provides the cathode buffer for electrophoresis and the right side provides for a capillary wash and spent polymer waste ejection functionality between injections. The Cathode Buffer Container Dx is a ready-to-use, disposable container with a radio frequency identification (RFID) tag incorporated into the label. The top of the Cathode Buffer Container Dx is heat sealed with a plastic film, which should be removed prior to direct installation on to the instrument. Each package includes four individual containers. The Septa Cathode Buffer Container Dx (sold separately) is used for sealing the Cathode Buffer Container Dx container.

This product is intended for in vitro diagnostic use on Applied Biosystems Dx genetic analyzers and is not validated for use with other systems or applications. Applied Biosystems Dx genetic analyzers are to be used by trained operators in the analysis of human DNA for the detection of fluorescently labeled DNA nucleotides by capillary electrophoresis. Refer to the appropriate Applied Biosystems Dx genetic analyzer user guide for further details.

The cathode buffer container (CBC) contains 1x running buffer to support all electrophoresis applications on the 3500 Dx series Genetic Analyzers.
For In Vitro Diagnostic Use Only
Specifications
For Use With (Equipment)3500xL Dx Genetic Analyzer, 3500 Dx Genetic Analyzer
Product TypeCathode Buffer Container
Quantity4 Ea.
Shipping ConditionWet Ice
Starting MaterialDNA, RNA
Unit SizeEach
Contents & Storage
Each package contains 4 buffer containers.
Store at 2 - 8°C.