N-TIPS pyrrole-3-boronic acid pinacol ester, 95%
N-TIPS pyrrole-3-boronic acid pinacol ester, 95%
N-TIPS pyrrole-3-boronic acid pinacol ester, 95%
Thermo Scientific Chemicals

N-TIPS pyrrole-3-boronic acid pinacol ester, 95%

CAS: 365564-11-0 | C19H36BNO2Si | 349.40 g/mol
数量:
1 g
5 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 441020010
価格(JPY)
-
数量:
1 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS365564-11-0
IUPAC Name3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1-[tris(propan-2-yl)silyl]-1H-pyrrole
Molecular FormulaC19H36BNO2Si
InChI KeyGWFIZBYDIHGZRJ-UHFFFAOYSA-N
SMILESCC(C)[Si](C(C)C)(C(C)C)N1C=CC(=C1)B1OC(C)(C)C(C)(C)O1
さらに表示
Appearance (Form)Powder or solid
GC>=94.0 %
Infrared spectrumConforms
Appearance (Color)White to brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only