3500 Dx Series 8-Tube Standard Retainer & Base Set
Applied Biosystems™

3500 Dx Series 8-Tube Standard Retainer & Base Set

This is an 8-tube retainer & base set (standard) designed for use with the 3500 Dx and 3500xL Dx genetic analyzers.
Have Questions?
Catalog NumberQuantity
44102304 Sets
Catalog number 4410230
Price (USD)
0.00
Each
Quantity:
4 Sets
Price (USD)
0.00
Each

This is a retainer and base set (standard) designed for septa retention in 8-tube strip format for use with the 3500 Dx and 3500xL Dx genetic analyzers.

Note: The 3500 Dx and 3500xL Dx genetic analyzers and consumables are for distribution and use in select countries only. Not for use in the USA.

  • This is an 8-tube retainer & base set (standard) designed for use with the 3500 Dx⁄3500xL Dx Genetic Analyzer.
  • Septa and plate base⁄retainer kits for 8 tube strip, 96 and 384-well plates used with the 3500 Dx Series Genetic Analyzers
  • The 3500 Dx Series Systems and consumables are for distribution and use in select countries only. Not for use in the USA.
For In Vitro Diagnostic Use Only
Specifications
For Use With (Equipment)3500xL Dx Genetic Analyzer, 3500 Dx Genetic Analyzer
Format8-tube Strip
Product TypeRetainer and Base Set
Quantity4 Sets
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
Ambient storage.
Each package contains 4 sets.