(S)-N-FMOC-α-Methylvaline, 98%, 98% ee, Thermo Scientific Chemicals
(S)-N-FMOC-α-Methylvaline, 98%, 98% ee, Thermo Scientific Chemicals
(S)-N-FMOC-α-Methylvaline, 98%, 98% ee, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(S)-N-FMOC-α-Methylvaline, 98%, 98% ee, Thermo Scientific Chemicals

CAS: 169566-81-8 | Molecular Formula: C21H23NO4 | Molecular Weight: 353.42 g/mol
Have Questions?
Catalog NumberQuantity
441155000500 mg
Catalog number 441155000
Price (USD)
-
Quantity:
500 mg
Request bulk or custom format
Chemical Identifiers
CAS169566-81-8
IUPAC Name2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-2,3-dimethylbutanoic acid
Molecular FormulaC21H23NO4
InChI KeyAWEZXIRZNQCCNN-NRFANRHFSA-N
SMILESCC(C)C(C)(NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Enantiomeric excess>=98.0 %
Residual solvents=<10.0 % MTBE
Specific optical rotation+7.5° to +9.5° (20°C, 589 nm) (c=1 in CHCl3)
Appearance (Color)White to light yellow
View more

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only