Which arrays can I use with the Invitrogen WT Expression Kit?
The Invitrogen WT Expression Kit is optimized for use with Affymetrix GeneChip Human, Mouse, and Rat 1.0 ST Arrays.
Search
View additional product information for WT Expression Kit - FAQs (4411973)
1 product FAQs found