cAMP-Screen™ Cyclic AMP Immunoassay System, 192 assays (2 x 96) - FAQs
cAMP-Screen™ Cyclic AMP Immunoassay System, 192 assays (2 x 96) - FAQs
View additional product information for cAMP-Screen™ Cyclic AMP Immunoassay System - FAQs (4412182, 4412183)
1 product FAQs found
What chemiluminescent detection assays do you offer for determining cyclic AMP levels in cell lysates?
We offer the cAMP-Screen and cAMP Screen Direct Cyclic AMP Immunoassay Systems for detection of cyclic AMP levels in cell lysates, see the products linked below:
cAMP-Screen Direct Cyclic AMP Immunoassay System (Cat. Nos. 4412186, 4412187)
cAMP-Screen Cyclic AMP Immunoassay System (Cat. Nos. 4412182, 4412183)