Not what you're looking for? Search all of our TaqMan® arrays for your gene or pathway of interest.
TaqMan™ Array Human MAPK Pathways
Applied Biosystems™

TaqMan™ Array Human MAPK Pathways

TaqMan™ Array Human MAPK Pathways
Have Questions?
Catalog NumberQuantity
44140931 standard plate
Catalog number 4414093
Price (USD)
273.00
Each
Quantity:
1 standard plate
Price (USD)
273.00
Each

These 96-well plates are pre-configured with the most appropriate TaqMan™ Gene Expression Assays for a specific biological process, pathway, or disease state. Each plate contains predefined assays and endogenous controls dried-down in the wells, ready for accurate assessment of an entire gene signature in one simple experiment.

The panel of assays in the TaqMan™ Array 96-well Human MAP Kinase Pathways Plate targets genes from the four major groups of mammalian MAPKs: 1) ERKs (Extracellular signal-Regulated Kinases); 2) p38 MAPKs; 3) JNKs (c-Jun NH2-terminal Kinases); and 4) ERK5 (or BMK cascades). Genes from the MAP2K (MAPKK), MAP3K (MAPKKK), and dual-specificity protein kinase MAPKK (DUSP) groups are also on the panel. Other groups of kinases in this panel include: cyclin dependent kinase (CDK), MAP kinase interacting serine⁄threonine kinase (MNK), p21 activated kinases (PAK), and ribosomal S6 kinases (RSK). Several MAPK targets including genes associated with signal transduction, transcription regulation, cell cycle regulation and cancer are also included.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
FormDried Down
IncludesTaqMan Assay for One 20 uL Reaction
Internal Probe ModificationMGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5')
Label or DyeFAM
Product LineTaqMan
Purification MethodStandard
Quantity1 standard plate
Reaction SpeedStandard
Research CategoryCell Cycle & Proliferation, Neurobiology, Signal Transduction, Inflammation, Development, Reproduction, Apoptosis, Oncology
TypeArray Plate
Format96-array Plate
SpeciesHuman
Unit SizeEach
Contents & Storage
Each TaqMan™ Array Plate well contains a single TaqMan™ Assay for one 20 μL reaction. The plates can be stored at room temperature or at 4°C.

Frequently asked questions (FAQs)

What is the minimum amount of TaqMan array plates I need to purchase to submit an order?

The minimum amount of TaqMan Array plates needed to submit an order is based on the plate type:
• Inventoried TaqMan array plates: 1 plate
• TaqMan Flexible Content panel array plates: 6 plates
• Custom TaqMan array plates: 6 plates
• Specialty TaqMan array plates: 10-20 plates (depends on assay format and configuration). Please contact our Specialty Plates team at Specialty_Plates@thermofisher.com for details.

Can our TaqMan Array Plates be run on non-Applied Biosystem real-time PCR instruments?

We recommend using our Plastics Selection Tool to verify which plates are compatible with your instrument model, https://www.thermofisher.com/us/en/home/life-science/pcr/pcr-plastics/plastics-selection-guide.html.

Based on our testing, the 96-well TaqMan Array Plates can be used on the following non-Applied Biosystem real-time PCR instruments:
• Standard (0.2mL) 96-well Plates: Bio-Rad IQ5
• Fast (0.1mL) Plates: Bio-Rad CFX Connect and Bio-Rad CFX96 Touch

How do I obtain TaqMan documentation files for my TaqMan array plates?

Beginning in April 2016, TaqMan documentation files for TaqMan Array Plates (assay information files (AIF), experimental setup files, and plate layout files) can be downloaded from the Thermo Fisher Scientific website at https://www.thermofisher.com/taqmanfiles. To obtain your TaqMan documentation files:
• Select the appropriate product type.
• Enter the information being requested for the product type.
• Fixed TaqMan array plates: Enter the Part number and Batch number
• TaqMan Flexible Content panel array plates/Custom TaqMan array plates: Enter the Sales Order number and Batch number
• Click Submit.