TaqMan™ Array Human IL1 Pathway - FAQs
TaqMan™ Array Human IL1 Pathway - FAQs
View additional product information for TaqMan™ Array Human IL1 Pathway - FAQs (4414142)
4 product FAQs found
Does TaqMan Array Human IL1 Pathway (Cat. No. 4414142) include the TaqMan PreAmp Master Mix (Cat. Nos. 4488593, 4391128)?
No. TaqMan Array Human IL1 Pathway (Cat. No. 4414142) does not include TaqMan PreAmp Master Mix (Cat. Nos. 4488593, 4391128). The required materials and equipment not supplied with TaqMan Gene Expression Assays and Array Plates can be found in the User Guide on page 9 and 10.
Find additional tips, troubleshooting help, and resources within our TaqMan Primers and Probes Support Center.
What is the minimum amount of TaqMan array plates I need to purchase to submit an order?
The minimum amount of TaqMan Array plates needed to submit an order is based on the plate type:
• Inventoried TaqMan array plates: 1 plate
• TaqMan Flexible Content panel array plates: 6 plates
• Custom TaqMan array plates: 6 plates
• Specialty TaqMan array plates: 10-20 plates (depends on assay format and configuration). Please contact our Specialty Plates team at Specialty_Plates@thermofisher.com for details.
Can our TaqMan Array Plates be run on non-Applied Biosystem real-time PCR instruments?
We recommend using our Plastics Selection Tool to verify which plates are compatible with your instrument model, https://www.thermofisher.com/us/en/home/life-science/pcr/pcr-plastics/plastics-selection-guide.html.
Based on our testing, the 96-well TaqMan Array Plates can be used on the following non-Applied Biosystem real-time PCR instruments:
• Standard (0.2mL) 96-well Plates: Bio-Rad IQ5
• Fast (0.1mL) Plates: Bio-Rad CFX Connect and Bio-Rad CFX96 Touch
How do I obtain TaqMan documentation files for my TaqMan array plates?
Beginning in April 2016, TaqMan documentation files for TaqMan Array Plates (assay information files (AIF), experimental setup files, and plate layout files) can be downloaded from the Thermo Fisher Scientific website at https://www.thermofisher.com/taqmanfiles. To obtain your TaqMan documentation files:
• Select the appropriate product type.
• Enter the information being requested for the product type.
• Fixed TaqMan array plates: Enter the Part number and Batch number