TaqMan™ 阵列人类 RAS 通路
Applied Biosystems™

TaqMan™ 阵列人类 RAS 通路

These 96-well plates are pre-configured with the most appropriate TaqMan® Gene Expression Assays for a specific biological process, pathway, or了解更多信息
Have Questions?
货号数量
44141761 standard plate
货号 4414176
价格(TWD)
14,280.00
飞享价
Ends: 30-Sep-2026
20,400.00
共减 6,120.00 (30%)
Each
数量:
1 standard plate
价格(TWD)
14,280.00
飞享价
Ends: 30-Sep-2026
20,400.00
共减 6,120.00 (30%)
Each
These 96-well plates are pre-configured with the most appropriate TaqMan® Gene Expression Assays for a specific biological process, pathway, or disease state. Each plate contains predefined assays and endogenous controls dried-down in the wells, ready for accurate assessment of an entire gene signature in one simple experiment. The TaqMan® Array Human RAS Pathway 96-well Plate contains 92 assays to genes associated with the RAS pathways and 4 assays to candidate endogenous control genes.
仅供科研使用。不可用于诊断程序。
规格
形式干燥
包括适用于一次 20 uL 反应的 TaqMan 检测
内部探针修改MGB(小沟结合物)(3')、不发出荧光淬灭剂 (3')、FAM (5')
标签或染料FAM
产品线TaqMan
数量1 standard plate
反应速度标准
研究类别细胞周期 & 增殖、神经生物学、信号转导、生物标记物相关通路、发育、免疫学、肿瘤学
类型阵列板
产品规格96 阵列板
种属
Unit SizeEach
内容与储存
每个 TaqMan™ 阵列板孔都包含一次 20 µL 反应所需的一种 TaqMan™ 检测。这些板可在室温或 4°C 下储存。

常见问题解答 (FAQ)

What is the minimum amount of TaqMan array plates I need to purchase to submit an order?

The minimum amount of TaqMan Array plates needed to submit an order is based on the plate type:
• Inventoried TaqMan array plates: 1 plate
• TaqMan Flexible Content panel array plates: 6 plates
• Custom TaqMan array plates: 6 plates
• Specialty TaqMan array plates: 10-20 plates (depends on assay format and configuration). Please contact our Specialty Plates team at Specialty_Plates@thermofisher.com for details.

Can our TaqMan Array Plates be run on non-Applied Biosystem real-time PCR instruments?

We recommend using our Plastics Selection Tool to verify which plates are compatible with your instrument model, https://www.thermofisher.com/us/en/home/life-science/pcr/pcr-plastics/plastics-selection-guide.html.

Based on our testing, the 96-well TaqMan Array Plates can be used on the following non-Applied Biosystem real-time PCR instruments:
• Standard (0.2mL) 96-well Plates: Bio-Rad IQ5
• Fast (0.1mL) Plates: Bio-Rad CFX Connect and Bio-Rad CFX96 Touch

How do I obtain TaqMan documentation files for my TaqMan array plates?

Beginning in April 2016, TaqMan documentation files for TaqMan Array Plates (assay information files (AIF), experimental setup files, and plate layout files) can be downloaded from the Thermo Fisher Scientific website at https://www.thermofisher.com/taqmanfiles. To obtain your TaqMan documentation files:
• Select the appropriate product type.
• Enter the information being requested for the product type.
• Fixed TaqMan array plates: Enter the Part number and Batch number
• TaqMan Flexible Content panel array plates/Custom TaqMan array plates: Enter the Sales Order number and Batch number
• Click Submit.