Dextran sulfate sodium salt, approx. M.W. 6,500 - 10,000
Dextran sulfate sodium salt, approx. M.W. 6,500 - 10,000
Dextran sulfate sodium salt, approx. M.W. 6,500 - 10,000
Thermo Scientific Chemicals

Dextran sulfate sodium salt, approx. M.W. 6,500 - 10,000

Dextran sulfate sodium salt, C9H11NO2, CAS Number-9011-18-1, americaine, anaesthesin, anesthesin, benzocaine, ethoform, ethyl aminobenzoate, ethyl p-aminobenzoate, norcaine, orthesin, parathesin, 25g, CHEBI:116735, BLFLLBZGZJTVJG-UHFFFAOYSA-N, ethyl 4-aminobenzoate, MFCD00081551, 165.192 | CAS: 9011-18-1 | (C6H7O5)m(C6H7O5)n | 40,000 g/mol
製品番号(カタログ番号) 441491000
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS9011-18-1
IUPAC NameDextran sulfate sodium salt
MDL NumberMFCD00081551
Specific optical rotation+75° to +105° (20°C, 589 nm) (6% in water)
Sulfur (S)16 to 20 %
Specific viscosity0.018 to 0.032 (1.0M in NaCl, 25°C)
Molecular weight6500 to 10000
Appearance (Color)White to light yellow to light brown
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only