TaqMan™ Array, Human Ligand Gated Ion Channels, Fast 96-well
These 96-well plates are pre-configured with the most appropriate TaqMan® Gene Expression Assays for a specific biological process, pathway, orRead more
Have Questions?
Catalog Number
Quantity
4418737
1 x 96-well plate
Catalog number 4418737
Price (USD)
273.00
Each
Quantity:
1 x 96-well plate
Price (USD)
273.00
Each
These 96-well plates are pre-configured with the most appropriate TaqMan® Gene Expression Assays for a specific biological process, pathway, or disease state. Each plate contains predefined assays and endogenous controls dried-down in the wells, ready for accurate assessment of an entire gene signature in one simple experiment. The panel of assays in the TaqMan® Array 96-well Human Ligand Gated Ion Channels Plate targets genes in the 3 superfamilies of the ligand-gated channels: 1) The Cys-loop receptors (GABAA receptor family, Glycine Receptor family, 5-HT Receptors⁄Serotonin, and Nicotinic acetylcholine receptor family); 2) The Ionotropic glutamate receptors (AMPA, Kainate, NMDA); and 3) The ATP-gated Channels (P2X receptors). Also in the panel are genes which encode Ion channel family members: Chloride channel, TRP (transient receptor potential) cation channels, Calcium Channel, and Sodium Channel. Additionally one gene from the Cyclic nucleotide-gated channel and Potassium gated ion channel are also present.
For Research Use Only. Not for use in diagnostic procedures.
Research CategoryCell Cycle & Proliferation, Signal Transduction, Inflammation
Format96-array Plate
SpeciesHuman
Unit SizeEach
Contents & Storage
Each TaqMan™ Array Plate well contains a single TaqMan™ Assay for one 10 μL reaction. The plates can be stored at room temperature or at 4°C.
Frequently asked questions (FAQs)
What mastermix can I use for running samples on TaqMan Gene Signature Array Plates?
We recommend that for standard (20 µL) TaqMan Array Plates, you use either the Universal Master Mix (I or II) or the Gene Expression MasterMix. For fast (10 µL) TaqMan Array Plates, use the Fast Universal Master Mix. The Fast Advanced Master Mix can also be used for either plate type, but we recommend following the protocol in this manual (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0025706_TaqManFastAdvMM_UG.pdf).
What is the minimum amount of TaqMan array plates I need to purchase to submit an order?
The minimum amount of TaqMan Array plates needed to submit an order is based on the plate type:
• Inventoried TaqMan array plates: 1 plate
• TaqMan Flexible Content panel array plates: 6 plates
• Custom TaqMan array plates: 6 plates
• Specialty TaqMan array plates: 10-20 plates (depends on assay format and configuration). Please contact our Specialty Plates team at Specialty_Plates@thermofisher.com for details.
Can our TaqMan Array Plates be run on non-Applied Biosystem real-time PCR instruments?
We recommend using our Plastics Selection Tool to verify which plates are compatible with your instrument model, https://www.thermofisher.com/us/en/home/life-science/pcr/pcr-plastics/plastics-selection-guide.html.
Based on our testing, the 96-well TaqMan Array Plates can be used on the following non-Applied Biosystem real-time PCR instruments:
• Standard (0.2mL) 96-well Plates: Bio-Rad IQ5
• Fast (0.1mL) Plates: Bio-Rad CFX Connect and Bio-Rad CFX96 Touch
How do I obtain TaqMan documentation files for my TaqMan array plates?
Beginning in April 2016, TaqMan documentation files for TaqMan Array Plates (assay information files (AIF), experimental setup files, and plate layout files) can be downloaded from the Thermo Fisher Scientific website at https://www.thermofisher.com/taqmanfiles. To obtain your TaqMan documentation files:
• Select the appropriate product type.
• Enter the information being requested for the product type.
• Fixed TaqMan array plates: Enter the Part number and Batch number
• TaqMan Flexible Content panel array plates/Custom TaqMan array plates: Enter the Sales Order number and Batch number
• Click Submit.