TaqMan™ Array, Human Nuclear Receptors, Fast 96-well - FAQs
TaqMan™ Array, Human Nuclear Receptors, Fast 96-well - FAQs
View additional product information for TaqMan™ Array, Human Nuclear Receptors, Fast 96-well - FAQs (4418753)
1 product FAQs found
What mastermix can I use for running samples on TaqMan Gene Signature Array Plates?
We recommend that for standard (20 µL) TaqMan Array Plates, you use either the Universal Master Mix (I or II) or the Gene Expression MasterMix. For fast (10 µL) TaqMan Array Plates, use the Fast Universal Master Mix. The Fast Advanced Master Mix can also be used for either plate type, but we recommend following the protocol in this manual (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0025706_TaqManFastAdvMM_UG.pdf).