TaqMan™ Array, Human Pancreatic Adenocarcinoma, Fast 96-well
Product Image
Applied Biosystems™

TaqMan™ Array, Human Pancreatic Adenocarcinoma, Fast 96-well

These 96-well plates are pre-configured with the most appropriate TaqMan® Gene Expression Assays for a specific biological process, pathway, or詳細を見る
製品番号(カタログ番号)数量
44188221 x 96-well plate
製品番号(カタログ番号) 4418822
価格(JPY)
26,900
Each
お問い合わせください ›
数量:
1 x 96-well plate
These 96-well plates are pre-configured with the most appropriate TaqMan® Gene Expression Assays for a specific biological process, pathway, or disease state. Each plate contains predefined assays and endogenous controls dried-down in the wells, ready for accurate assessment of an entire gene signature in one simple experiment. The TaqMan® Array Human Pancreatic Adenocarinoma 96-well Plate contains 92 assays to pancreatic adenocarinoma associated genes and 4 assays to candidate endogenous control genes.
研究用途にのみご使用ください。診断目的には使用できません。
仕様
形状乾燥
内容TaqMan Assay for One 10 uL Reaction
内部プローブの変更MGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5')
標識または色素FAM
製品ラインTaqMan
数量1 x 96-well plate
反応速度高速
研究カテゴリーImmunology, Signal Transduction, Biomarker-Related Pathway(s), Extracellular Matrix & Cell Adhesion, Oncology
タイプArray Plate
フォーマット96-array Plate
Human
Unit SizeEach
組成および保存条件
Each TaqMan™ Array Plate well contains a single TaqMan™ Assay for one 10 μL reaction. The plates can be stored at room temperature or at 4°C.

よくあるご質問(FAQ)

TaqMan® Gene Signature Array Platesを使用する場合は、どのマスターミックスを使用すれば良いでしょうか?

Standard (20μL) のTaqMan® Array Platesには、Universal Master Mix (I または II)、若しくは Gene Expression Master Mixが使用可能です。Fast (10μL) のTaqMan® Array Platesには、Fast Universal Master Mixが使用可能です。Fast Advanced Master Mixは、どちらのプレートタイプにも使用可能ですが、 推奨プロトコルに従いご使用ください。

What mastermix can I use for running samples on TaqMan Gene Signature Array Plates?

We recommend that for standard (20 µL) TaqMan Array Plates, you use either the Universal Master Mix (I or II) or the Gene Expression MasterMix. For fast (10 µL) TaqMan Array Plates, use the Fast Universal Master Mix. The Fast Advanced Master Mix can also be used for either plate type, but we recommend following the protocol in this manual (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0025706_TaqManFastAdvMM_UG.pdf).

What is the minimum amount of TaqMan array plates I need to purchase to submit an order?

The minimum amount of TaqMan Array plates needed to submit an order is based on the plate type:
• Inventoried TaqMan array plates: 1 plate
• TaqMan Flexible Content panel array plates: 6 plates
• Custom TaqMan array plates: 6 plates
• Specialty TaqMan array plates: 10-20 plates (depends on assay format and configuration). Please contact our Specialty Plates team at Specialty_Plates@thermofisher.com for details.

Can our TaqMan Array Plates be run on non-Applied Biosystem real-time PCR instruments?

We recommend using our Plastics Selection Tool to verify which plates are compatible with your instrument model, https://www.thermofisher.com/us/en/home/life-science/pcr/pcr-plastics/plastics-selection-guide.html.

Based on our testing, the 96-well TaqMan Array Plates can be used on the following non-Applied Biosystem real-time PCR instruments:
• Standard (0.2mL) 96-well Plates: Bio-Rad IQ5
• Fast (0.1mL) Plates: Bio-Rad CFX Connect and Bio-Rad CFX96 Touch

How do I obtain TaqMan documentation files for my TaqMan array plates?

Beginning in April 2016, TaqMan documentation files for TaqMan Array Plates (assay information files (AIF), experimental setup files, and plate layout files) can be downloaded from the Thermo Fisher Scientific website at https://www.thermofisher.com/taqmanfiles. To obtain your TaqMan documentation files:
• Select the appropriate product type.
• Enter the information being requested for the product type.
• Fixed TaqMan array plates: Enter the Part number and Batch number
• TaqMan Flexible Content panel array plates/Custom TaqMan array plates: Enter the Sales Order number and Batch number
• Click Submit.