Finnpipette™ Dispenser
Thermo Scientific™

Finnpipette™ Dispenser

Dose liquids from reagent bottles with dispensers that offer superior chemical and thermal resistance, and an ergonomic design that prevents work fatigue.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)
442117010.0 to 60.0 mL
44211200.2 to 1.0 mL
44211300.4 to 2.0 mL
44211401.0 to 5.0 mL
44211502.0 to 10.0 mL
44211605.0 to 30.0 mL
Catalog number 4421170
Price (USD)
1,091.70
Online exclusive
1,237.00
Save 145.30
Each
Volume (Metric):
10.0 to 60.0 mL
Price (USD)
1,091.70
Online exclusive
1,237.00
Save 145.30
Each
Dose liquids from reagent bottles with Thermo Scientific™ Finnpipette™ Dispensers.

Features:

  • Excellent chemical and thermal resistance
  • Fully autoclavable†
  • Exact self-locking system in the volume setting
  • Dispensing head cap prevents dripping
  • Ergonomic design prevents work fatigue
  • Threaded suction hose for bubble-free suction

Note:

Limitations include HF, liquids that attack Halar, FEP or Hastelloy suspensions because solid particles might block the valves.

Warranty: 1 year

Note: Not for use on hot plates.
Specifications
Certifications/ComplianceIndividual calibration certificate with serial number
DescriptionRange: 10-60mL, ±360µL
Precision120 μL
Channels1
Product TypeAnalog Bottletop Dispenser
Volume (Metric)10.0 to 60.0 mL
Unit SizeEach