2,3-Dimethoxy-5-methyl-1,4-benzoquinone, 98%
2,3-Dimethoxy-5-methyl-1,4-benzoquinone, 98%
2,3-Dimethoxy-5-methyl-1,4-benzoquinone, 98%
Thermo Scientific Chemicals

2,3-Dimethoxy-5-methyl-1,4-benzoquinone, 98%

CAS: 605-94-7 | C9H10O4 | 182.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 442200010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS605-94-7
IUPAC Name2,3-dimethoxy-5-methylcyclohexa-2,5-diene-1,4-dione
Molecular FormulaC9H10O4
InChI KeyUIXPTCZPFCVOQF-UHFFFAOYSA-N
SMILESCOC1=C(OC)C(=O)C(C)=CC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange to red
Appearance (Form)Crystalline powder or needles
Melting point58°C to 62°C
HPLC>=97.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only